Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160222564-160222835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8
Entrez Gene ID
50717
Gene Name
DDB1 and CUL4 associated factor 8
Gene Aliases
GAN2, H326, WDR42A
UniGene
Hs.632447
Gene Chromosome Location
Chr.1: 160215715-160262560 on Build GRCh38
Transcripts
NM_015726.3 NR_028103.1 NR_028104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249753 c.1862G>A p.C621Y Chr.1: 160222691-160222691 on Build GRCh38
5731293 c.1790A>G p.Y597C Chr.1: 160222763-160222763 on Build GRCh38
4464442 c.1334C>T p.P445L Chr.1: 160222757-160222757 on Build GRCh38
6891460 c.1358G>A p.G453D Chr.1: 160222733-160222733 on Build GRCh38
6058686 c.1438G>A p.V480M Chr.1: 160222653-160222653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144537625 Chr.1: 160222683-160222683 on Build GRCh38
rs755879191 Chr.1: 160222661-160222661 on Build GRCh38
rs759585222 Chr.1: 160222803-160222803 on Build GRCh38
rs773491214 Chr.1: 160222762-160222762 on Build GRCh38
rs371339023 Chr.1: 160222624-160222624 on Build GRCh38
rs369037918 Chr.1: 160222627-160222627 on Build GRCh38
rs896102130 Chr.1: 160222617-160222617 on Build GRCh38
rs1037242127 Chr.1: 160222594-160222594 on Build GRCh38
rs746284391 Chr.1: 160222687-160222687 on Build GRCh38
rs780377544 Chr.1: 160222697-160222697 on Build GRCh38
rs754486094 Chr.1: 160222632-160222632 on Build GRCh38
rs774313299 Chr.1: 160222639-160222640 on Build GRCh38
rs767163915 Chr.1: 160222653-160222653 on Build GRCh38
rs112012061 Chr.1: 160222797-160222797 on Build GRCh38
rs758804943 Chr.1: 160222693-160222693 on Build GRCh38
rs578202487 Chr.1: 160222814-160222814 on Build GRCh38
rs772942213 Chr.1: 160222621-160222621 on Build GRCh38
rs1029528953 Chr.1: 160222666-160222666 on Build GRCh38
rs200095819 Chr.1: 160222725-160222725 on Build GRCh38
rs762065942 Chr.1: 160222609-160222609 on Build GRCh38
rs769981566 Chr.1: 160222744-160222744 on Build GRCh38
rs35813537 Chr.1: 160222735-160222736 on Build GRCh38
rs1032128621 Chr.1: 160222794-160222794 on Build GRCh38
rs762947432 Chr.1: 160222622-160222622 on Build GRCh38
rs964120621 Chr.1: 160222648-160222648 on Build GRCh38
rs748358744 Chr.1: 160222735-160222735 on Build GRCh38
rs767112541 Chr.1: 160222809-160222809 on Build GRCh38
rs776744486 Chr.1: 160222717-160222717 on Build GRCh38
rs553627512 Chr.1: 160222790-160222790 on Build GRCh38
rs766216059 Chr.1: 160222774-160222774 on Build GRCh38
rs764996740 Chr.1: 160222615-160222615 on Build GRCh38
rs142684645 Chr.1: 160222710-160222710 on Build GRCh38
rs956730220 Chr.1: 160222668-160222668 on Build GRCh38
rs751525040 Chr.1: 160222630-160222630 on Build GRCh38
rs752366102 Chr.1: 160222654-160222654 on Build GRCh38
rs768693156 Chr.1: 160222711-160222711 on Build GRCh38
rs762593998 Chr.1: 160222773-160222773 on Build GRCh38
rs776795592 Chr.1: 160222601-160222601 on Build GRCh38

View Full Product Details