Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63959955-63960216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMSD
Entrez Gene ID
284293
Gene Name
histocompatibility minor serpin domain containing
Gene Aliases
ACC-6, ACC6, C18orf53, HSMD-v
UniGene
Hs.721604
Gene Chromosome Location
Chr.18: 63949259-63969329 on Build GRCh38
Transcripts
NM_001123366.1 XM_011525930.2 XM_017025710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3527157 c.236C>T p.S79F Chr.18: 63960171-63960171 on Build GRCh38
7371353 c.280G>T p.V94L Chr.18: 63960215-63960215 on Build GRCh38
4895132 c.275A>G p.D92G Chr.18: 63960210-63960210 on Build GRCh38
4831952 c.230C>T p.T77I Chr.18: 63960165-63960165 on Build GRCh38
3144715 c.261G>A p.T87T Chr.18: 63960196-63960196 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768256115 Chr.18: 63959999-63959999 on Build GRCh38
rs750865752 Chr.18: 63960148-63960148 on Build GRCh38
rs114535947 Chr.18: 63960018-63960018 on Build GRCh38
rs754190361 Chr.18: 63960035-63960035 on Build GRCh38
rs767938646 Chr.18: 63960108-63960109 on Build GRCh38
rs147152052 Chr.18: 63959990-63959990 on Build GRCh38
rs12959740 Chr.18: 63960066-63960066 on Build GRCh38
rs1042548724 Chr.18: 63960123-63960123 on Build GRCh38
rs376302490 Chr.18: 63960172-63960172 on Build GRCh38
rs373069173 Chr.18: 63960168-63960168 on Build GRCh38
rs376770809 Chr.18: 63960118-63960118 on Build GRCh38
rs756949228 Chr.18: 63960139-63960139 on Build GRCh38
rs777425464 Chr.18: 63960126-63960126 on Build GRCh38
rs1014393266 Chr.18: 63959979-63959979 on Build GRCh38
rs372180749 Chr.18: 63960111-63960111 on Build GRCh38
rs539361759 Chr.18: 63960188-63960188 on Build GRCh38
rs943641863 Chr.18: 63960077-63960077 on Build GRCh38
rs369452873 Chr.18: 63960166-63960166 on Build GRCh38
rs751207567 Chr.18: 63960131-63960131 on Build GRCh38
rs745915253 Chr.18: 63960154-63960154 on Build GRCh38
rs780749620 Chr.18: 63960146-63960146 on Build GRCh38
rs749662389 Chr.18: 63960152-63960152 on Build GRCh38
rs765214334 Chr.18: 63960109-63960109 on Build GRCh38
rs779840921 Chr.18: 63960033-63960033 on Build GRCh38
rs989298299 Chr.18: 63960078-63960078 on Build GRCh38
rs1033414104 Chr.18: 63960029-63960029 on Build GRCh38
rs577020675 Chr.18: 63960084-63960084 on Build GRCh38
rs773298780 Chr.18: 63960181-63960181 on Build GRCh38
rs1000220067 Chr.18: 63960027-63960027 on Build GRCh38
rs760788825 Chr.18: 63960186-63960186 on Build GRCh38
rs976241288 Chr.18: 63960097-63960097 on Build GRCh38
rs769895614 Chr.18: 63960187-63960187 on Build GRCh38
rs748662951 Chr.18: 63960171-63960171 on Build GRCh38

View Full Product Details