Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241685694-241685965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTYMK
Entrez Gene ID
1841
Gene Name
deoxythymidylate kinase
Gene Aliases
CDC8, PP3731, TMPK, TYMK
UniGene
Hs.471873
Gene Chromosome Location
Chr.2: 241675742-241686968 on Build GRCh38
Transcripts
NM_001165031.1 NM_001320902.1 NM_001320903.1 NM_001320904.1 NM_001320905.1 NM_012145.3 NR_033255.1 NR_135492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6449640 c.200C>T p.S67L Chr.2: 241685808-241685808 on Build GRCh38
418781 c.149G>A p.G50D Chr.2: 241685859-241685859 on Build GRCh38
227713 c.226C>T p.R76C Chr.2: 241685782-241685782 on Build GRCh38
4679643 c.215T>G p.F72C Chr.2: 241685793-241685793 on Build GRCh38
4533942 c.202G>A p.V68M Chr.2: 241685806-241685806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868274031 Chr.2: 241685869-241685869 on Build GRCh38
rs370211112 Chr.2: 241685851-241685851 on Build GRCh38
rs952397128 Chr.2: 241685843-241685843 on Build GRCh38
rs1051965767 Chr.2: 241685858-241685858 on Build GRCh38
rs984301835 Chr.2: 241685849-241685849 on Build GRCh38
rs561956735 Chr.2: 241685752-241685752 on Build GRCh38
rs775258374 Chr.2: 241685835-241685835 on Build GRCh38
rs778053080 Chr.2: 241685761-241685761 on Build GRCh38
rs761416404 Chr.2: 241685861-241685861 on Build GRCh38
rs34209418 Chr.2: 241685791-241685791 on Build GRCh38
rs975331316 Chr.2: 241685808-241685808 on Build GRCh38
rs759330928 Chr.2: 241685825-241685825 on Build GRCh38
rs765737654 Chr.2: 241685891-241685891 on Build GRCh38
rs868650343 Chr.2: 241685859-241685859 on Build GRCh38
rs775235055 Chr.2: 241685888-241685889 on Build GRCh38
rs753092156 Chr.2: 241685909-241685909 on Build GRCh38
rs772165781 Chr.2: 241685886-241685889 on Build GRCh38
rs755384797 Chr.2: 241685731-241685731 on Build GRCh38
rs760649892 Chr.2: 241685893-241685894 on Build GRCh38
rs773823928 Chr.2: 241685860-241685860 on Build GRCh38
rs942012561 Chr.2: 241685821-241685821 on Build GRCh38
rs940226709 Chr.2: 241685943-241685943 on Build GRCh38
rs1028417590 Chr.2: 241685810-241685810 on Build GRCh38
rs754427846 Chr.2: 241685868-241685868 on Build GRCh38
rs574574617 Chr.2: 241685807-241685807 on Build GRCh38
rs766880836 Chr.2: 241685863-241685863 on Build GRCh38
rs998687388 Chr.2: 241685726-241685726 on Build GRCh38
rs769831932 Chr.2: 241685925-241685927 on Build GRCh38
rs759059953 Chr.2: 241685745-241685745 on Build GRCh38
rs1019577933 Chr.2: 241685798-241685798 on Build GRCh38
rs747138010 Chr.2: 241685797-241685797 on Build GRCh38
rs1009994393 Chr.2: 241685924-241685924 on Build GRCh38
rs1039432925 Chr.2: 241685841-241685841 on Build GRCh38
rs769630524 Chr.2: 241685831-241685831 on Build GRCh38
rs763713666 Chr.2: 241685856-241685856 on Build GRCh38
rs755536782 Chr.2: 241685873-241685873 on Build GRCh38
rs201918916 Chr.2: 241685726-241685727 on Build GRCh38
rs748447686 Chr.2: 241685751-241685751 on Build GRCh38

View Full Product Details