Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55785258-55785531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX4
Entrez Gene ID
54514
Gene Name
DEAD-box helicase 4
Gene Aliases
VASA
UniGene
Hs.223581
Gene Chromosome Location
Chr.5: 55737994-55817146 on Build GRCh38
Transcripts
NM_001142549.1 NM_001166533.1 NM_001166534.1 NM_024415.2 XM_011543495.2 XM_011543497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398321 c.686C>A p.S229* Chr.5: 55785459-55785459 on Build GRCh38
3344453 c.260G>A p.S87N Chr.5: 55785480-55785480 on Build GRCh38
6398320 c.239C>A p.S80* Chr.5: 55785459-55785459 on Build GRCh38
1695795 c.718C>T p.Q240* Chr.5: 55785491-55785491 on Build GRCh38
1620403 c.262A>C p.S88R Chr.5: 55785482-55785482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759895893 Chr.5: 55785479-55785479 on Build GRCh38
rs771217180 Chr.5: 55785398-55785398 on Build GRCh38
rs922214074 Chr.5: 55785362-55785362 on Build GRCh38
rs750392307 Chr.5: 55785393-55785393 on Build GRCh38
rs144187220 Chr.5: 55785339-55785339 on Build GRCh38
rs772582408 Chr.5: 55785335-55785335 on Build GRCh38
rs763799913 Chr.5: 55785348-55785348 on Build GRCh38
rs774449788 Chr.5: 55785306-55785306 on Build GRCh38
rs374255122 Chr.5: 55785336-55785336 on Build GRCh38
rs771435941 Chr.5: 55785460-55785460 on Build GRCh38
rs768894231 Chr.5: 55785296-55785296 on Build GRCh38
rs761835357 Chr.5: 55785309-55785309 on Build GRCh38
rs778994474 Chr.5: 55785423-55785423 on Build GRCh38
rs926495013 Chr.5: 55785334-55785334 on Build GRCh38
rs773268378 Chr.5: 55785444-55785444 on Build GRCh38
rs751135942 Chr.5: 55785355-55785355 on Build GRCh38
rs759732020 Chr.5: 55785466-55785468 on Build GRCh38
rs748231812 Chr.5: 55785426-55785426 on Build GRCh38
rs772207237 Chr.5: 55785430-55785430 on Build GRCh38
rs749568181 Chr.5: 55785354-55785354 on Build GRCh38
rs528171437 Chr.5: 55785403-55785403 on Build GRCh38
rs779756501 Chr.5: 55785419-55785419 on Build GRCh38
rs778211924 Chr.5: 55785285-55785288 on Build GRCh38
rs776888797 Chr.5: 55785468-55785468 on Build GRCh38
rs773179185 Chr.5: 55785297-55785297 on Build GRCh38
rs201596382 Chr.5: 55785346-55785346 on Build GRCh38
rs34200022 Chr.5: 55785459-55785459 on Build GRCh38
rs781275293 Chr.5: 55785386-55785386 on Build GRCh38
rs754207044 Chr.5: 55785340-55785340 on Build GRCh38
rs763851557 Chr.5: 55785489-55785489 on Build GRCh38
rs199749889 Chr.5: 55785488-55785488 on Build GRCh38
rs756765558 Chr.5: 55785381-55785381 on Build GRCh38
rs774596476 Chr.5: 55785407-55785408 on Build GRCh38
rs749073333 Chr.5: 55785421-55785421 on Build GRCh38

View Full Product Details