Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46371667-46371902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO2
Entrez Gene ID
63916
Gene Name
engulfment and cell motility 2
Gene Aliases
CED-12, CED12, Ced-12A, ELMO-2
UniGene
Hs.210469
Gene Chromosome Location
Chr.20: 46366045-46407051 on Build GRCh38
Transcripts
NM_001318253.1 NM_133171.4 NM_182764.2 XM_005260496.3 XM_005260498.3 XM_005260499.3 XM_005260500.3 XM_005260501.4 XM_006723854.3 XM_017028009.1 XM_017028010.1 XM_017028011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4099001 c.1532G>A p.R511H Chr.20: 46371854-46371854 on Build GRCh38
724377 c.1499G>T p.R500L Chr.20: 46371887-46371887 on Build GRCh38
6378397 c.1535A>T p.Q512L Chr.20: 46371851-46371851 on Build GRCh38
6004413 c.1531C>T p.R511C Chr.20: 46371855-46371855 on Build GRCh38
6441079 c.1540G>A p.E514K Chr.20: 46371846-46371846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760241662 Chr.20: 46371696-46371696 on Build GRCh38
rs747728081 Chr.20: 46371864-46371864 on Build GRCh38
rs778085258 Chr.20: 46371811-46371811 on Build GRCh38
rs376458042 Chr.20: 46371816-46371816 on Build GRCh38
rs961394374 Chr.20: 46371753-46371753 on Build GRCh38
rs913674833 Chr.20: 46371707-46371707 on Build GRCh38
rs769284976 Chr.20: 46371871-46371871 on Build GRCh38
rs546819827 Chr.20: 46371778-46371778 on Build GRCh38
rs775583783 Chr.20: 46371694-46371694 on Build GRCh38
rs570435889 Chr.20: 46371699-46371699 on Build GRCh38
rs776049108 Chr.20: 46371862-46371862 on Build GRCh38
rs772866712 Chr.20: 46371872-46371872 on Build GRCh38
rs926280664 Chr.20: 46371732-46371732 on Build GRCh38
rs370879690 Chr.20: 46371779-46371779 on Build GRCh38
rs972450025 Chr.20: 46371760-46371760 on Build GRCh38
rs993357068 Chr.20: 46371823-46371823 on Build GRCh38
rs748841493 Chr.20: 46371710-46371712 on Build GRCh38
rs763234588 Chr.20: 46371877-46371877 on Build GRCh38
rs774822992 Chr.20: 46371879-46371879 on Build GRCh38
rs757981591 Chr.20: 46371719-46371719 on Build GRCh38
rs756327149 Chr.20: 46371801-46371801 on Build GRCh38
rs1016697465 Chr.20: 46371723-46371723 on Build GRCh38
rs766811805 Chr.20: 46371781-46371781 on Build GRCh38
rs762231531 Chr.20: 46371773-46371774 on Build GRCh38
rs149080790 Chr.20: 46371687-46371687 on Build GRCh38
rs767962684 Chr.20: 46371790-46371791 on Build GRCh38
rs752937635 Chr.20: 46371795-46371795 on Build GRCh38
rs747002155 Chr.20: 46371850-46371850 on Build GRCh38
rs767551212 Chr.20: 46371788-46371788 on Build GRCh38
rs752065089 Chr.20: 46371782-46371782 on Build GRCh38
rs538756608 Chr.20: 46371820-46371820 on Build GRCh38
rs774615848 Chr.20: 46371733-46371733 on Build GRCh38
rs780136305 Chr.20: 46371847-46371847 on Build GRCh38
rs761421278 Chr.20: 46371730-46371730 on Build GRCh38
rs745607349 Chr.20: 46371815-46371815 on Build GRCh38
rs143362697 Chr.20: 46371726-46371727 on Build GRCh38
rs766793261 Chr.20: 46371878-46371878 on Build GRCh38
rs985676110 Chr.20: 46371810-46371810 on Build GRCh38
rs773794464 Chr.20: 46371791-46371791 on Build GRCh38
rs2297052 Chr.20: 46371783-46371783 on Build GRCh38
rs768190847 Chr.20: 46371860-46371860 on Build GRCh38
rs199553318 Chr.20: 46371718-46371718 on Build GRCh38
rs746456458 Chr.20: 46371706-46371706 on Build GRCh38

View Full Product Details