Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134358963-134359236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L2
Entrez Gene ID
89944
Gene Name
galactosidase beta 1 like 2
Gene Aliases
MST114, MSTP114
UniGene
Hs.436178
Gene Chromosome Location
Chr.11: 134331874-134376324 on Build GRCh38
Transcripts
NM_138342.3 XM_006718937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3979418 c.658G>T p.E220* Chr.11: 134359066-134359066 on Build GRCh38
5448128 c.710T>C p.L237P Chr.11: 134359118-134359118 on Build GRCh38
5499580 c.680T>C p.L227P Chr.11: 134359088-134359088 on Build GRCh38
6486030 c.695A>T p.D232V Chr.11: 134359103-134359103 on Build GRCh38
242533 c.673G>T p.V225L Chr.11: 134359081-134359081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200721051 Chr.11: 134359073-134359073 on Build GRCh38
rs557827058 Chr.11: 134359163-134359163 on Build GRCh38
rs1040338074 Chr.11: 134359206-134359206 on Build GRCh38
rs954062257 Chr.11: 134359116-134359116 on Build GRCh38
rs985395411 Chr.11: 134359129-134359129 on Build GRCh38
rs986588597 Chr.11: 134359176-134359176 on Build GRCh38
rs745359050 Chr.11: 134359139-134359139 on Build GRCh38
rs766502573 Chr.11: 134359058-134359058 on Build GRCh38
rs201831404 Chr.11: 134359175-134359175 on Build GRCh38
rs771042266 Chr.11: 134359103-134359105 on Build GRCh38
rs756502548 Chr.11: 134359185-134359185 on Build GRCh38
rs750626221 Chr.11: 134359051-134359051 on Build GRCh38
rs763140288 Chr.11: 134359052-134359052 on Build GRCh38
rs561963338 Chr.11: 134359199-134359199 on Build GRCh38
rs1000679935 Chr.11: 134359037-134359037 on Build GRCh38
rs377260743 Chr.11: 134359033-134359033 on Build GRCh38
rs914390586 Chr.11: 134359170-134359170 on Build GRCh38
rs59983932 Chr.11: 134359160-134359160 on Build GRCh38
rs775251567 Chr.11: 134359155-134359155 on Build GRCh38
rs199851534 Chr.11: 134359123-134359123 on Build GRCh38
rs776661511 Chr.11: 134359203-134359203 on Build GRCh38
rs972464187 Chr.11: 134359156-134359156 on Build GRCh38
rs540388475 Chr.11: 134359178-134359178 on Build GRCh38
rs771889275 Chr.11: 134359025-134359025 on Build GRCh38
rs773575745 Chr.11: 134359164-134359164 on Build GRCh38
rs778450536 Chr.11: 134359138-134359138 on Build GRCh38
rs1032249946 Chr.11: 134359039-134359039 on Build GRCh38
rs951868896 Chr.11: 134359115-134359115 on Build GRCh38
rs765528931 Chr.11: 134359034-134359034 on Build GRCh38
rs990023617 Chr.11: 134359169-134359169 on Build GRCh38
rs777491794 Chr.11: 134359104-134359104 on Build GRCh38
rs764507172 Chr.11: 134359187-134359187 on Build GRCh38
rs370824928 Chr.11: 134359132-134359132 on Build GRCh38
rs765693289 Chr.11: 134359146-134359146 on Build GRCh38
rs768086946 Chr.11: 134359179-134359179 on Build GRCh38
rs774412808 Chr.11: 134359114-134359114 on Build GRCh38
rs759535595 Chr.11: 134359015-134359015 on Build GRCh38
rs772087679 Chr.11: 134359141-134359141 on Build GRCh38
rs753084313 Chr.11: 134359182-134359182 on Build GRCh38
rs370291095 Chr.11: 134359165-134359165 on Build GRCh38
rs756382082 Chr.11: 134359103-134359103 on Build GRCh38
rs575572908 Chr.11: 134359130-134359130 on Build GRCh38
rs775404545 Chr.11: 134359027-134359027 on Build GRCh38
rs1040579306 Chr.11: 134358988-134358988 on Build GRCh38
rs150906098 Chr.11: 134359127-134359127 on Build GRCh38
rs755604496 Chr.11: 134359072-134359072 on Build GRCh38
rs753133595 Chr.11: 134359093-134359093 on Build GRCh38
rs774274119 Chr.11: 134359014-134359014 on Build GRCh38
rs147438844 Chr.11: 134358987-134358987 on Build GRCh38
rs944678826 Chr.11: 134359205-134359205 on Build GRCh38
rs529259686 Chr.11: 134359212-134359212 on Build GRCh38
rs751688239 Chr.11: 134359063-134359063 on Build GRCh38

View Full Product Details