Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45777459-45777720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMPK
Entrez Gene ID
1760
Gene Name
dystrophia myotonica protein kinase
Gene Aliases
DM, DM1, DM1PK, DMK, MDPK, MT-PK
UniGene
Hs.631596
Gene Chromosome Location
Chr.19: 45769709-45782557 on Build GRCh38
Transcripts
NM_001081560.2 NM_001081562.2 NM_001081563.2 NM_001288764.1 NM_001288765.1 NM_001288766.1 NM_004409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390113 c.898C>T p.P300S Chr.19: 45777575-45777575 on Build GRCh38
3990118 c.998C>T p.T333I Chr.19: 45777505-45777505 on Build GRCh38
7095686 c.834C>G p.F278L Chr.19: 45777715-45777715 on Build GRCh38
6645665 c.945G>A p.G315G Chr.19: 45777558-45777558 on Build GRCh38
3990119 c.968C>T p.T323I Chr.19: 45777505-45777505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962168090 Chr.19: 45777674-45777674 on Build GRCh38
rs780885997 Chr.19: 45777683-45777683 on Build GRCh38
rs374895405 Chr.19: 45777654-45777654 on Build GRCh38
rs773706825 Chr.19: 45777563-45777563 on Build GRCh38
rs201714957 Chr.19: 45777541-45777541 on Build GRCh38
rs750253660 Chr.19: 45777655-45777655 on Build GRCh38
rs528187884 Chr.19: 45777612-45777612 on Build GRCh38
rs762230984 Chr.19: 45777485-45777485 on Build GRCh38
rs757185658 Chr.19: 45777584-45777584 on Build GRCh38
rs139724884 Chr.19: 45777514-45777514 on Build GRCh38
rs751225267 Chr.19: 45777662-45777662 on Build GRCh38
rs999563080 Chr.19: 45777581-45777581 on Build GRCh38
rs761285661 Chr.19: 45777480-45777480 on Build GRCh38
rs370770361 Chr.19: 45777616-45777616 on Build GRCh38
rs200058479 Chr.19: 45777564-45777564 on Build GRCh38
rs149803658 Chr.19: 45777580-45777580 on Build GRCh38
rs570705950 Chr.19: 45777698-45777698 on Build GRCh38
rs905980461 Chr.19: 45777570-45777570 on Build GRCh38
rs747639886 Chr.19: 45777689-45777689 on Build GRCh38
rs749666645 Chr.19: 45777699-45777699 on Build GRCh38
rs140158944 Chr.19: 45777633-45777633 on Build GRCh38
rs767991260 Chr.19: 45777607-45777607 on Build GRCh38
rs771214913 Chr.19: 45777626-45777626 on Build GRCh38
rs766485074 Chr.19: 45777573-45777573 on Build GRCh38
rs750614096 Chr.19: 45777490-45777490 on Build GRCh38
rs759774926 Chr.19: 45777527-45777527 on Build GRCh38
rs146157132 Chr.19: 45777525-45777525 on Build GRCh38
rs745640651 Chr.19: 45777594-45777594 on Build GRCh38
rs751679078 Chr.19: 45777574-45777574 on Build GRCh38
rs776266757 Chr.19: 45777611-45777611 on Build GRCh38
rs779534958 Chr.19: 45777602-45777602 on Build GRCh38
rs771592769 Chr.19: 45777529-45777529 on Build GRCh38
rs756122292 Chr.19: 45777511-45777511 on Build GRCh38
rs372132101 Chr.19: 45777697-45777697 on Build GRCh38
rs770389641 Chr.19: 45777617-45777617 on Build GRCh38
rs554967270 Chr.19: 45777637-45777637 on Build GRCh38
rs141629958 Chr.19: 45777659-45777659 on Build GRCh38
rs746534953 Chr.19: 45777605-45777605 on Build GRCh38
rs771259599 Chr.19: 45777700-45777700 on Build GRCh38
rs147815859 Chr.19: 45777585-45777585 on Build GRCh38
rs763177539 Chr.19: 45777625-45777625 on Build GRCh38
rs749089111 Chr.19: 45777519-45777519 on Build GRCh38
rs766547162 Chr.19: 45777495-45777495 on Build GRCh38
rs141319926 Chr.19: 45777691-45777691 on Build GRCh38
rs762280354 Chr.19: 45777542-45777542 on Build GRCh38
rs201008625 Chr.19: 45777632-45777632 on Build GRCh38
rs774774759 Chr.19: 45777701-45777701 on Build GRCh38
rs777077770 Chr.19: 45777484-45777484 on Build GRCh38
rs752743750 Chr.19: 45777640-45777640 on Build GRCh38
rs754651761 Chr.19: 45777675-45777675 on Build GRCh38
rs778669160 Chr.19: 45777526-45777526 on Build GRCh38
rs765558854 Chr.19: 45777489-45777489 on Build GRCh38
rs781155031 Chr.19: 45777620-45777620 on Build GRCh38
rs758065350 Chr.19: 45777658-45777658 on Build GRCh38
rs764488056 Chr.19: 45777481-45777481 on Build GRCh38
rs769239379 Chr.19: 45777540-45777540 on Build GRCh38
rs555191515 Chr.19: 45777493-45777493 on Build GRCh38
rs545769272 Chr.19: 45777578-45777578 on Build GRCh38
rs765754023 Chr.19: 45777558-45777558 on Build GRCh38
rs563294621 Chr.19: 45777533-45777533 on Build GRCh38
rs758092645 Chr.19: 45777601-45777601 on Build GRCh38
rs150617093 Chr.19: 45777510-45777510 on Build GRCh38
rs773760078 Chr.19: 45777622-45777622 on Build GRCh38

View Full Product Details