Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133636329-133636596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBH
Entrez Gene ID
1621
Gene Name
dopamine beta-hydroxylase
Gene Aliases
DBM
UniGene
Hs.591890
Gene Chromosome Location
Chr.9: 133636363-133659344 on Build GRCh38
Transcripts
NM_000787.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
310481 c.35G>T p.G12V Chr.9: 133636406-133636406 on Build GRCh38
3952451 c.22G>A p.A8T Chr.9: 133636393-133636393 on Build GRCh38
6114907 c.104C>A p.A35D Chr.9: 133636475-133636475 on Build GRCh38
3952452 c.45G>A p.M15I Chr.9: 133636416-133636416 on Build GRCh38
3655334 c.222C>T p.F74F Chr.9: 133636593-133636593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764517520 Chr.9: 133636471-133636471 on Build GRCh38
rs758863303 Chr.9: 133636378-133636378 on Build GRCh38
rs368414868 Chr.9: 133636459-133636459 on Build GRCh38
rs13306306 Chr.9: 133636417-133636417 on Build GRCh38
rs762193644 Chr.9: 133636542-133636542 on Build GRCh38
rs778274517 Chr.9: 133636384-133636384 on Build GRCh38
rs778828919 Chr.9: 133636572-133636572 on Build GRCh38
rs200561151 Chr.9: 133636540-133636540 on Build GRCh38
rs775268917 Chr.9: 133636426-133636426 on Build GRCh38
rs111514228 Chr.9: 133636387-133636387 on Build GRCh38
rs755302704 Chr.9: 133636568-133636568 on Build GRCh38
rs750763532 Chr.9: 133636544-133636544 on Build GRCh38
rs76856960 Chr.9: 133636447-133636447 on Build GRCh38
rs770187471 Chr.9: 133636402-133636402 on Build GRCh38
rs745685569 Chr.9: 133636517-133636517 on Build GRCh38
rs746901698 Chr.9: 133636501-133636501 on Build GRCh38
rs756815809 Chr.9: 133636443-133636443 on Build GRCh38
rs930083685 Chr.9: 133636462-133636462 on Build GRCh38
rs547488212 Chr.9: 133636496-133636496 on Build GRCh38
rs769637610 Chr.9: 133636417-133636418 on Build GRCh38
rs766920830 Chr.9: 133636557-133636557 on Build GRCh38
rs944060307 Chr.9: 133636508-133636508 on Build GRCh38
rs139591190 Chr.9: 133636536-133636536 on Build GRCh38
rs770621845 Chr.9: 133636433-133636433 on Build GRCh38
rs762462999 Chr.9: 133636460-133636460 on Build GRCh38
rs760403889 Chr.9: 133636349-133636349 on Build GRCh38
rs1028535785 Chr.9: 133636507-133636507 on Build GRCh38
rs199920333 Chr.9: 133636377-133636377 on Build GRCh38
rs781667738 Chr.9: 133636388-133636388 on Build GRCh38
rs755891058 Chr.9: 133636490-133636490 on Build GRCh38
rs78562123 Chr.9: 133636519-133636519 on Build GRCh38
rs200378894 Chr.9: 133636491-133636491 on Build GRCh38
rs143544421 Chr.9: 133636477-133636477 on Build GRCh38
rs533854976 Chr.9: 133636541-133636541 on Build GRCh38
rs984418612 Chr.9: 133636549-133636549 on Build GRCh38
rs374469015 Chr.9: 133636358-133636358 on Build GRCh38
rs770943084 Chr.9: 133636506-133636506 on Build GRCh38
rs373639922 Chr.9: 133636396-133636396 on Build GRCh38
rs562042962 Chr.9: 133636487-133636487 on Build GRCh38
rs374987238 Chr.9: 133636575-133636575 on Build GRCh38
rs769378064 Chr.9: 133636525-133636525 on Build GRCh38
rs751081253 Chr.9: 133636415-133636415 on Build GRCh38
rs140025171 Chr.9: 133636476-133636476 on Build GRCh38
rs779109570 Chr.9: 133636499-133636499 on Build GRCh38
rs754101732 Chr.9: 133636562-133636562 on Build GRCh38
rs746061650 Chr.9: 133636393-133636393 on Build GRCh38
rs146820764 Chr.9: 133636498-133636498 on Build GRCh38
rs564703488 Chr.9: 133636437-133636437 on Build GRCh38
rs868839518 Chr.9: 133636367-133636367 on Build GRCh38
rs753429940 Chr.9: 133636370-133636370 on Build GRCh38
rs963604579 Chr.9: 133636418-133636418 on Build GRCh38
rs775127844 Chr.9: 133636535-133636535 on Build GRCh38
rs5318 Chr.9: 133636405-133636405 on Build GRCh38
rs767747480 Chr.9: 133636480-133636480 on Build GRCh38
rs776251904 Chr.9: 133636510-133636510 on Build GRCh38
rs959727837 Chr.9: 133636394-133636394 on Build GRCh38
rs756555116 Chr.9: 133636554-133636554 on Build GRCh38
rs780246471 Chr.9: 133636404-133636404 on Build GRCh38
rs201797804 Chr.9: 133636408-133636408 on Build GRCh38
rs768096757 Chr.9: 133636543-133636543 on Build GRCh38
rs776539329 Chr.9: 133636435-133636435 on Build GRCh38

View Full Product Details