Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74436622-74436855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69C
Entrez Gene ID
125704
Gene Name
family with sequence similarity 69 member C
Gene Aliases
C18orf51
UniGene
Hs.371690
Gene Chromosome Location
Chr.18: 74432575-74464646 on Build GRCh38
Transcripts
NM_001044369.2 XM_011525816.2 XM_017025551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972642 c.190A>T p.K64* Chr.18: 74436674-74436674 on Build GRCh38
5972641 c.1087A>T p.K363* Chr.18: 74436674-74436674 on Build GRCh38
1389695 c.167G>A p.R56H Chr.18: 74436697-74436697 on Build GRCh38
6226956 c.156C>G p.D52E Chr.18: 74436708-74436708 on Build GRCh38
3146265 c.237G>A p.A79A Chr.18: 74436627-74436627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770311448 Chr.18: 74436743-74436743 on Build GRCh38
rs746634956 Chr.18: 74436640-74436640 on Build GRCh38
rs905615914 Chr.18: 74436822-74436822 on Build GRCh38
rs748356608 Chr.18: 74436664-74436664 on Build GRCh38
rs949768863 Chr.18: 74436773-74436773 on Build GRCh38
rs984432430 Chr.18: 74436690-74436690 on Build GRCh38
rs201783677 Chr.18: 74436662-74436662 on Build GRCh38
rs1045371863 Chr.18: 74436822-74436822 on Build GRCh38
rs1053427697 Chr.18: 74436682-74436682 on Build GRCh38
rs775925637 Chr.18: 74436678-74436679 on Build GRCh38
rs200796961 Chr.18: 74436683-74436683 on Build GRCh38
rs561393471 Chr.18: 74436749-74436749 on Build GRCh38
rs749381624 Chr.18: 74436748-74436748 on Build GRCh38
rs373904492 Chr.18: 74436708-74436708 on Build GRCh38
rs762130860 Chr.18: 74436666-74436666 on Build GRCh38
rs775918221 Chr.18: 74436747-74436747 on Build GRCh38
rs762428869 Chr.18: 74436832-74436832 on Build GRCh38
rs538254406 Chr.18: 74436786-74436786 on Build GRCh38
rs778627012 Chr.18: 74436694-74436694 on Build GRCh38
rs767358198 Chr.18: 74436675-74436675 on Build GRCh38
rs371974579 Chr.18: 74436697-74436697 on Build GRCh38
rs754507772 Chr.18: 74436689-74436689 on Build GRCh38
rs758060528 Chr.18: 74436698-74436698 on Build GRCh38
rs766264139 Chr.18: 74436684-74436684 on Build GRCh38
rs112746285 Chr.18: 74436833-74436833 on Build GRCh38
rs11875335 Chr.18: 74436663-74436663 on Build GRCh38
rs753368911 Chr.18: 74436688-74436688 on Build GRCh38
rs750262375 Chr.18: 74436681-74436681 on Build GRCh38
rs146689233 Chr.18: 74436735-74436735 on Build GRCh38

View Full Product Details