Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 128277805-128278078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBN2
Entrez Gene ID
2201
Gene Name
fibrillin 2
Gene Aliases
CCA, DA9, EOMD
UniGene
Hs.519294
Gene Chromosome Location
Chr.5: 128257908-128538358 on Build GRCh38
Transcripts
NM_001999.3 XM_017009228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6490259 c.7412C>A p.S2471* Chr.5: 128277939-128277939 on Build GRCh38
149904 c.7417C>T p.R2473* Chr.5: 128277934-128277934 on Build GRCh38
6302581 c.7442C>G p.T2481S Chr.5: 128277909-128277909 on Build GRCh38
3365797 c.7355A>C p.E2452A Chr.5: 128277996-128277996 on Build GRCh38
7275863 c.7402A>G p.T2468A Chr.5: 128277949-128277949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547811908 Chr.5: 128278046-128278046 on Build GRCh38
rs576679175 Chr.5: 128277883-128277883 on Build GRCh38
rs375822966 Chr.5: 128278028-128278028 on Build GRCh38
rs28763925 Chr.5: 128277933-128277933 on Build GRCh38
rs371525539 Chr.5: 128277833-128277833 on Build GRCh38
rs940249059 Chr.5: 128277843-128277843 on Build GRCh38
rs145397241 Chr.5: 128278050-128278050 on Build GRCh38
rs781735118 Chr.5: 128278027-128278027 on Build GRCh38
rs751962930 Chr.5: 128277987-128277987 on Build GRCh38
rs1000679210 Chr.5: 128277831-128277831 on Build GRCh38
rs199639432 Chr.5: 128278043-128278043 on Build GRCh38
rs774331714 Chr.5: 128277949-128277949 on Build GRCh38
rs369559870 Chr.5: 128277875-128277875 on Build GRCh38
rs374577668 Chr.5: 128277840-128277840 on Build GRCh38
rs908807863 Chr.5: 128277842-128277842 on Build GRCh38
rs1016476735 Chr.5: 128277882-128277882 on Build GRCh38
rs527729655 Chr.5: 128278044-128278044 on Build GRCh38
rs768488202 Chr.5: 128277937-128277937 on Build GRCh38
rs747531309 Chr.5: 128277902-128277902 on Build GRCh38
rs778405069 Chr.5: 128277897-128277897 on Build GRCh38
rs963985988 Chr.5: 128277917-128277917 on Build GRCh38
rs28763924 Chr.5: 128277859-128277859 on Build GRCh38
rs1013655545 Chr.5: 128277858-128277858 on Build GRCh38
rs753059448 Chr.5: 128277876-128277876 on Build GRCh38
rs778349551 Chr.5: 128277985-128277985 on Build GRCh38
rs375739609 Chr.5: 128278022-128278022 on Build GRCh38
rs749843181 Chr.5: 128277962-128277962 on Build GRCh38
rs200998513 Chr.5: 128277874-128277874 on Build GRCh38
rs147102633 Chr.5: 128277971-128277971 on Build GRCh38
rs745974408 Chr.5: 128277918-128277918 on Build GRCh38
rs753596079 Chr.5: 128277976-128277976 on Build GRCh38
rs758823011 Chr.5: 128277981-128277981 on Build GRCh38
rs373360372 Chr.5: 128277860-128277860 on Build GRCh38
rs1053601562 Chr.5: 128277942-128277942 on Build GRCh38
rs759743264 Chr.5: 128277836-128277836 on Build GRCh38
rs373370945 Chr.5: 128277955-128277955 on Build GRCh38
rs771043310 Chr.5: 128277911-128277911 on Build GRCh38
rs769956474 Chr.5: 128277932-128277932 on Build GRCh38
rs73784562 Chr.5: 128277849-128277849 on Build GRCh38
rs863223546 Chr.5: 128277946-128277946 on Build GRCh38
rs950688484 Chr.5: 128277872-128277872 on Build GRCh38
rs562508529 Chr.5: 128277951-128277951 on Build GRCh38
rs776223919 Chr.5: 128278022-128278023 on Build GRCh38
rs1009522430 Chr.5: 128277959-128277959 on Build GRCh38
rs374605626 Chr.5: 128278029-128278029 on Build GRCh38
rs1028505966 Chr.5: 128278034-128278034 on Build GRCh38
rs863223587 Chr.5: 128277982-128277982 on Build GRCh38
rs576501460 Chr.5: 128277966-128277966 on Build GRCh38
rs750081283 Chr.5: 128277873-128277873 on Build GRCh38
rs781066710 Chr.5: 128277914-128277914 on Build GRCh38
rs762760752 Chr.5: 128277844-128277844 on Build GRCh38
rs766993329 Chr.5: 128277863-128277863 on Build GRCh38
rs1019541548 Chr.5: 128277969-128277969 on Build GRCh38

View Full Product Details