Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57992874-57993119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A2
Entrez Gene ID
8854
Gene Name
aldehyde dehydrogenase 1 family member A2
Gene Aliases
RALDH(II), RALDH2, RALDH2-T
UniGene
Hs.643455
Gene Chromosome Location
Chr.15: 57953424-58065923 on Build GRCh38
Transcripts
NM_001206897.1 NM_003888.3 NM_170696.2 NM_170697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404681 c.631C>T p.P211S Chr.15: 57992998-57992998 on Build GRCh38
4660999 c.670G>A p.A224T Chr.15: 57992959-57992959 on Build GRCh38
6401771 c.642A>C p.Q214H Chr.15: 57992987-57992987 on Build GRCh38
1373754 c.653G>A p.S218N Chr.15: 57992976-57992976 on Build GRCh38
3502739 c.567C>T p.P189P Chr.15: 57993062-57993062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746584588 Chr.15: 57993028-57993028 on Build GRCh38
rs776426483 Chr.15: 57992934-57992934 on Build GRCh38
rs368422746 Chr.15: 57993059-57993059 on Build GRCh38
rs781200515 Chr.15: 57993041-57993041 on Build GRCh38
rs116291924 Chr.15: 57992957-57992957 on Build GRCh38
rs757661279 Chr.15: 57992992-57992992 on Build GRCh38
rs368104935 Chr.15: 57992927-57992927 on Build GRCh38
rs760824618 Chr.15: 57992973-57992973 on Build GRCh38
rs931784193 Chr.15: 57992913-57992913 on Build GRCh38
rs769420073 Chr.15: 57992956-57992956 on Build GRCh38
rs747977737 Chr.15: 57992947-57992947 on Build GRCh38
rs758095172 Chr.15: 57993018-57993018 on Build GRCh38
rs200511796 Chr.15: 57992930-57992930 on Build GRCh38
rs201462764 Chr.15: 57993081-57993081 on Build GRCh38
rs767821847 Chr.15: 57992959-57992959 on Build GRCh38
rs764707551 Chr.15: 57993081-57993082 on Build GRCh38
rs760423034 Chr.15: 57992908-57992908 on Build GRCh38
rs138763874 Chr.15: 57992926-57992926 on Build GRCh38
rs757560541 Chr.15: 57992922-57992922 on Build GRCh38
rs372191854 Chr.15: 57993079-57993079 on Build GRCh38
rs141814790 Chr.15: 57992933-57992933 on Build GRCh38
rs769664771 Chr.15: 57993068-57993068 on Build GRCh38
rs764488032 Chr.15: 57992903-57992903 on Build GRCh38
rs531974410 Chr.15: 57992921-57992921 on Build GRCh38
rs754288575 Chr.15: 57992905-57992905 on Build GRCh38
rs4474631 Chr.15: 57992981-57992981 on Build GRCh38
rs35740908 Chr.15: 57992904-57992905 on Build GRCh38
rs765478305 Chr.15: 57993009-57993009 on Build GRCh38
rs375479963 Chr.15: 57992991-57992991 on Build GRCh38
rs777472837 Chr.15: 57993076-57993076 on Build GRCh38
rs779663083 Chr.15: 57993021-57993021 on Build GRCh38
rs397897208 Chr.15: 57992906-57992907 on Build GRCh38
rs759794887 Chr.15: 57992958-57992958 on Build GRCh38
rs371271832 Chr.15: 57993087-57993087 on Build GRCh38
rs754414994 Chr.15: 57993036-57993036 on Build GRCh38
rs764730133 Chr.15: 57992976-57992976 on Build GRCh38
rs566704871 Chr.15: 57992969-57992969 on Build GRCh38
rs746496383 Chr.15: 57992932-57992932 on Build GRCh38
rs750755375 Chr.15: 57993013-57993013 on Build GRCh38
rs775761595 Chr.15: 57992965-57992965 on Build GRCh38

View Full Product Details