Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62413815-62414040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP8NL
Entrez Gene ID
140893
Gene Name
RBBP8 N-terminal like
Gene Aliases
C20orf151
UniGene
Hs.180374
Gene Chromosome Location
Chr.20: 62410237-62427573 on Build GRCh38
Transcripts
NM_080833.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568434 c.1460G>T p.S487I Chr.20: 62413891-62413891 on Build GRCh38
3389880 c.1404C>T p.A468A Chr.20: 62413947-62413947 on Build GRCh38
6526048 c.1335G>T p.S445S Chr.20: 62414016-62414016 on Build GRCh38
5966750 c.1509G>T p.Q503H Chr.20: 62413842-62413842 on Build GRCh38
6423167 c.1412A>G p.H471R Chr.20: 62413939-62413939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777291306 Chr.20: 62414017-62414017 on Build GRCh38
rs780657453 Chr.20: 62413999-62413999 on Build GRCh38
rs377275739 Chr.20: 62413908-62413908 on Build GRCh38
rs768490738 Chr.20: 62413863-62413863 on Build GRCh38
rs771453716 Chr.20: 62413851-62413851 on Build GRCh38
rs924597688 Chr.20: 62414016-62414016 on Build GRCh38
rs202197217 Chr.20: 62413895-62413895 on Build GRCh38
rs950066696 Chr.20: 62413995-62413995 on Build GRCh38
rs769554211 Chr.20: 62414014-62414014 on Build GRCh38
rs765508964 Chr.20: 62413984-62413984 on Build GRCh38
rs375826789 Chr.20: 62413947-62413947 on Build GRCh38
rs746646870 Chr.20: 62413862-62413862 on Build GRCh38
rs6061520 Chr.20: 62413958-62413958 on Build GRCh38
rs746341470 Chr.20: 62413934-62413934 on Build GRCh38
rs903248303 Chr.20: 62413887-62413887 on Build GRCh38
rs757657322 Chr.20: 62413841-62413841 on Build GRCh38
rs368768680 Chr.20: 62413915-62413915 on Build GRCh38
rs778102712 Chr.20: 62413837-62413837 on Build GRCh38
rs779376394 Chr.20: 62413927-62413927 on Build GRCh38
rs749591643 Chr.20: 62413838-62413838 on Build GRCh38
rs754504409 Chr.20: 62414005-62414005 on Build GRCh38
rs934531122 Chr.20: 62413893-62413893 on Build GRCh38
rs754032472 Chr.20: 62413979-62413979 on Build GRCh38
rs749050845 Chr.20: 62414022-62414022 on Build GRCh38
rs769450666 Chr.20: 62413964-62413964 on Build GRCh38
rs752837907 Chr.20: 62413921-62413921 on Build GRCh38
rs780896548 Chr.20: 62414006-62414006 on Build GRCh38
rs774288427 Chr.20: 62413976-62413976 on Build GRCh38
rs757524019 Chr.20: 62413926-62413926 on Build GRCh38
rs779242362 Chr.20: 62413842-62413842 on Build GRCh38
rs773114589 Chr.20: 62413973-62413973 on Build GRCh38
rs759002620 Chr.20: 62413938-62413938 on Build GRCh38
rs898664800 Chr.20: 62413992-62413992 on Build GRCh38
rs767479432 Chr.20: 62413919-62413919 on Build GRCh38
rs756186645 Chr.20: 62413922-62413922 on Build GRCh38
rs76137593 Chr.20: 62413993-62413993 on Build GRCh38
rs776382035 Chr.20: 62413948-62413948 on Build GRCh38
rs189749375 Chr.20: 62413907-62413907 on Build GRCh38
rs746420402 Chr.20: 62413844-62413844 on Build GRCh38
rs747739276 Chr.20: 62414009-62414009 on Build GRCh38
rs370742035 Chr.20: 62413894-62413894 on Build GRCh38
rs752143359 Chr.20: 62414000-62414000 on Build GRCh38
rs758836984 Chr.20: 62413847-62413847 on Build GRCh38
rs544985213 Chr.20: 62413975-62413975 on Build GRCh38
rs867257247 Chr.20: 62413906-62413906 on Build GRCh38
rs892958341 Chr.20: 62413899-62413899 on Build GRCh38
rs764192442 Chr.20: 62413978-62413978 on Build GRCh38
rs757549106 Chr.20: 62413982-62413982 on Build GRCh38
rs775116137 Chr.20: 62413857-62413857 on Build GRCh38
rs1015368739 Chr.20: 62413928-62413928 on Build GRCh38
rs762790428 Chr.20: 62413974-62413974 on Build GRCh38
rs148498049 Chr.20: 62413925-62413925 on Build GRCh38
rs779708200 Chr.20: 62413946-62413946 on Build GRCh38
rs760700893 Chr.20: 62413977-62413977 on Build GRCh38
rs758695091 Chr.20: 62413994-62413994 on Build GRCh38
rs764247509 Chr.20: 62413924-62413924 on Build GRCh38
rs950015310 Chr.20: 62413852-62413852 on Build GRCh38

View Full Product Details