Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144357400-144357673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
UniGene
Hs.12271
Gene Chromosome Location
Chr.8: 144355428-144358523 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
366712 c.575G>T p.R192L Chr.8: 144357628-144357628 on Build GRCh38
1191130 c.612delG p.W204fs*1 Chr.8: 144357466-144357466 on Build GRCh38
1190808 c.613delA p.K205fs*8 Chr.8: 144357465-144357465 on Build GRCh38
4770070 c.569C>A p.P190H Chr.8: 144357634-144357634 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371785275 Chr.8: 144357607-144357607 on Build GRCh38
rs934697642 Chr.8: 144357568-144357568 on Build GRCh38
rs781956203 Chr.8: 144357627-144357627 on Build GRCh38
rs978864497 Chr.8: 144357526-144357526 on Build GRCh38
rs782689018 Chr.8: 144357516-144357516 on Build GRCh38
rs782233238 Chr.8: 144357525-144357525 on Build GRCh38
rs191369551 Chr.8: 144357616-144357616 on Build GRCh38
rs782740194 Chr.8: 144357581-144357581 on Build GRCh38
rs781900166 Chr.8: 144357591-144357591 on Build GRCh38
rs782264436 Chr.8: 144357613-144357613 on Build GRCh38
rs782788981 Chr.8: 144357589-144357589 on Build GRCh38
rs368491938 Chr.8: 144357572-144357572 on Build GRCh38
rs781815063 Chr.8: 144357490-144357490 on Build GRCh38
rs376118339 Chr.8: 144357635-144357635 on Build GRCh38
rs782111638 Chr.8: 144357463-144357463 on Build GRCh38
rs376158451 Chr.8: 144357457-144357457 on Build GRCh38
rs781838985 Chr.8: 144357466-144357466 on Build GRCh38
rs782347231 Chr.8: 144357456-144357456 on Build GRCh38
rs374417005 Chr.8: 144357556-144357556 on Build GRCh38
rs187124016 Chr.8: 144357603-144357603 on Build GRCh38
rs782449940 Chr.8: 144357596-144357596 on Build GRCh38
rs782182756 Chr.8: 144357619-144357619 on Build GRCh38
rs781919874 Chr.8: 144357425-144357425 on Build GRCh38
rs926012786 Chr.8: 144357554-144357554 on Build GRCh38
rs782151544 Chr.8: 144357535-144357535 on Build GRCh38
rs782316664 Chr.8: 144357450-144357450 on Build GRCh38
rs782644488 Chr.8: 144357620-144357620 on Build GRCh38
rs782572770 Chr.8: 144357600-144357600 on Build GRCh38
rs782069792 Chr.8: 144357549-144357549 on Build GRCh38
rs782472216 Chr.8: 144357505-144357505 on Build GRCh38
rs557063176 Chr.8: 144357509-144357509 on Build GRCh38
rs781851282 Chr.8: 144357582-144357582 on Build GRCh38
rs367644123 Chr.8: 144357520-144357520 on Build GRCh38
rs756548422 Chr.8: 144357631-144357631 on Build GRCh38
rs1004998551 Chr.8: 144357637-144357637 on Build GRCh38
rs374712587 Chr.8: 144357514-144357514 on Build GRCh38
rs114506049 Chr.8: 144357505-144357505 on Build GRCh38
rs782329010 Chr.8: 144357623-144357623 on Build GRCh38
rs782241678 Chr.8: 144357639-144357639 on Build GRCh38
rs781864548 Chr.8: 144357510-144357510 on Build GRCh38
rs1035386434 Chr.8: 144357424-144357424 on Build GRCh38
rs200802955 Chr.8: 144357576-144357576 on Build GRCh38
rs558884638 Chr.8: 144357583-144357583 on Build GRCh38
rs576723144 Chr.8: 144357533-144357533 on Build GRCh38
rs782387059 Chr.8: 144357629-144357629 on Build GRCh38
rs782424635 Chr.8: 144357542-144357542 on Build GRCh38
rs782599808 Chr.8: 144357521-144357521 on Build GRCh38
rs782640771 Chr.8: 144357611-144357611 on Build GRCh38
rs782034686 Chr.8: 144357524-144357524 on Build GRCh38
rs886185736 Chr.8: 144357620-144357620 on Build GRCh38
rs782697334 Chr.8: 144357648-144357648 on Build GRCh38
rs368413712 Chr.8: 144357478-144357478 on Build GRCh38
rs1034697377 Chr.8: 144357652-144357652 on Build GRCh38
rs782696648 Chr.8: 144357617-144357617 on Build GRCh38
rs368478706 Chr.8: 144357650-144357650 on Build GRCh38
rs781923203 Chr.8: 144357544-144357544 on Build GRCh38
rs568018357 Chr.8: 144357448-144357448 on Build GRCh38

View Full Product Details