Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46853901-46854140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRF5
Entrez Gene ID
221395
Gene Name
adhesion G protein-coupled receptor F5
Gene Aliases
GPR116, KPG_001
UniGene
Hs.362806
Gene Chromosome Location
Chr.6: 46852505-46954954 on Build GRCh38
Transcripts
NM_001098518.1 NM_015234.4 XM_005248892.2 XM_005248893.3 XM_005248894.3 XM_005248895.3 XM_011514353.2 XM_011514354.2 XM_017010418.1 XM_017010419.1 XM_017010420.1 XM_017010421.1 XM_017010422.1 XM_017010423.1 XM_017010424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874556 c.3970A>C p.N1324H Chr.6: 46854063-46854063 on Build GRCh38
3628012 c.3986A>G p.E1329G Chr.6: 46854047-46854047 on Build GRCh38
6284127 c.3964A>G p.T1322A Chr.6: 46854069-46854069 on Build GRCh38
3874555 c.3976T>C p.S1326P Chr.6: 46854057-46854057 on Build GRCh38
1079945 c.4034T>A p.L1345H Chr.6: 46853999-46853999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751120085 Chr.6: 46854075-46854075 on Build GRCh38
rs140284327 Chr.6: 46854068-46854068 on Build GRCh38
rs201720856 Chr.6: 46853961-46853961 on Build GRCh38
rs779661188 Chr.6: 46854061-46854063 on Build GRCh38
rs769508037 Chr.6: 46854008-46854008 on Build GRCh38
rs767827928 Chr.6: 46854044-46854044 on Build GRCh38
rs763001357 Chr.6: 46854017-46854017 on Build GRCh38
rs1007007834 Chr.6: 46854100-46854100 on Build GRCh38
rs938454426 Chr.6: 46854042-46854042 on Build GRCh38
rs531431396 Chr.6: 46854051-46854051 on Build GRCh38
rs755884882 Chr.6: 46854098-46854098 on Build GRCh38
rs779551843 Chr.6: 46853991-46853991 on Build GRCh38
rs747779867 Chr.6: 46854082-46854082 on Build GRCh38
rs780858421 Chr.6: 46854080-46854080 on Build GRCh38
rs759816951 Chr.6: 46854034-46854034 on Build GRCh38
rs780829581 Chr.6: 46854005-46854005 on Build GRCh38
rs117945872 Chr.6: 46853970-46853970 on Build GRCh38
rs547640157 Chr.6: 46854090-46854090 on Build GRCh38
rs745981189 Chr.6: 46854119-46854119 on Build GRCh38
rs374150291 Chr.6: 46853977-46853977 on Build GRCh38
rs368830573 Chr.6: 46854021-46854021 on Build GRCh38
rs147148177 Chr.6: 46854032-46854032 on Build GRCh38
rs746596600 Chr.6: 46853996-46853996 on Build GRCh38
rs1056995384 Chr.6: 46854053-46854053 on Build GRCh38
rs753792039 Chr.6: 46853979-46853979 on Build GRCh38
rs369973653 Chr.6: 46853960-46853960 on Build GRCh38
rs562739561 Chr.6: 46853930-46853930 on Build GRCh38
rs147835950 Chr.6: 46853946-46853947 on Build GRCh38
rs368868523 Chr.6: 46854078-46854078 on Build GRCh38
rs552188864 Chr.6: 46854013-46854013 on Build GRCh38
rs768357396 Chr.6: 46854004-46854004 on Build GRCh38
rs373405244 Chr.6: 46853971-46853971 on Build GRCh38
rs761056955 Chr.6: 46854046-46854046 on Build GRCh38
rs549945311 Chr.6: 46854067-46854067 on Build GRCh38
rs775683084 Chr.6: 46854045-46854045 on Build GRCh38
rs888111419 Chr.6: 46854099-46854099 on Build GRCh38
rs376634437 Chr.6: 46853984-46853984 on Build GRCh38
rs769586233 Chr.6: 46853966-46853967 on Build GRCh38
rs778381574 Chr.6: 46853981-46853981 on Build GRCh38
rs774548607 Chr.6: 46854111-46854111 on Build GRCh38
rs201165369 Chr.6: 46853946-46853946 on Build GRCh38
rs769821080 Chr.6: 46853955-46853955 on Build GRCh38
rs773134483 Chr.6: 46853956-46853956 on Build GRCh38
rs371219762 Chr.6: 46853987-46853987 on Build GRCh38
rs144111839 Chr.6: 46853989-46853989 on Build GRCh38
rs771053869 Chr.6: 46854109-46854109 on Build GRCh38
rs754285982 Chr.6: 46854055-46854055 on Build GRCh38
rs565850477 Chr.6: 46854105-46854105 on Build GRCh38
rs374789760 Chr.6: 46854112-46854112 on Build GRCh38
rs375764569 Chr.6: 46853965-46853965 on Build GRCh38

View Full Product Details