Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40367350-40367623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISP2
Entrez Gene ID
85455
Gene Name
dispatched RND transporter family member 2
Gene Aliases
DISPB, HsT16908, LINC00594
UniGene
Hs.355645
Gene Chromosome Location
Chr.15: 40354359-40378755 on Build GRCh38
Transcripts
NM_033510.2 XM_011522125.2 XM_017022687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077224 c.1435G>A p.E479K Chr.15: 40367547-40367547 on Build GRCh38
3500860 c.1416G>A p.M472I Chr.15: 40367528-40367528 on Build GRCh38
2269351 c.1376C>T p.P459L Chr.15: 40367488-40367488 on Build GRCh38
3500861 c.1474C>T p.P492S Chr.15: 40367586-40367586 on Build GRCh38
2269352 c.1381G>A p.G461R Chr.15: 40367493-40367493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771114862 Chr.15: 40367539-40367539 on Build GRCh38
rs762084264 Chr.15: 40367448-40367448 on Build GRCh38
rs768059804 Chr.15: 40367489-40367489 on Build GRCh38
rs749976957 Chr.15: 40367506-40367506 on Build GRCh38
rs773259271 Chr.15: 40367525-40367525 on Build GRCh38
rs75971826 Chr.15: 40367446-40367446 on Build GRCh38
rs143680406 Chr.15: 40367542-40367542 on Build GRCh38
rs150246885 Chr.15: 40367556-40367556 on Build GRCh38
rs190907713 Chr.15: 40367569-40367569 on Build GRCh38
rs753096222 Chr.15: 40367492-40367492 on Build GRCh38
rs374719365 Chr.15: 40367476-40367476 on Build GRCh38
rs759792631 Chr.15: 40367588-40367588 on Build GRCh38
rs377416042 Chr.15: 40367379-40367379 on Build GRCh38
rs150131260 Chr.15: 40367520-40367520 on Build GRCh38
rs764960634 Chr.15: 40367565-40367565 on Build GRCh38
rs745345275 Chr.15: 40367400-40367400 on Build GRCh38
rs749399952 Chr.15: 40367529-40367529 on Build GRCh38
rs145288960 Chr.15: 40367503-40367503 on Build GRCh38
rs939282505 Chr.15: 40367404-40367404 on Build GRCh38
rs370777054 Chr.15: 40367424-40367424 on Build GRCh38
rs915195900 Chr.15: 40367518-40367518 on Build GRCh38
rs779975608 Chr.15: 40367510-40367510 on Build GRCh38
rs756981544 Chr.15: 40367500-40367500 on Build GRCh38
rs760905436 Chr.15: 40367564-40367564 on Build GRCh38
rs1017146165 Chr.15: 40367412-40367412 on Build GRCh38
rs200056671 Chr.15: 40367516-40367516 on Build GRCh38
rs566140400 Chr.15: 40367421-40367421 on Build GRCh38
rs753346991 Chr.15: 40367564-40367564 on Build GRCh38
rs754951771 Chr.15: 40367576-40367576 on Build GRCh38
rs375390991 Chr.15: 40367441-40367441 on Build GRCh38
rs865811298 Chr.15: 40367438-40367438 on Build GRCh38
rs772083845 Chr.15: 40367418-40367418 on Build GRCh38
rs962819493 Chr.15: 40367419-40367419 on Build GRCh38
rs746825288 Chr.15: 40367436-40367436 on Build GRCh38
rs942713637 Chr.15: 40367583-40367583 on Build GRCh38
rs138813855 Chr.15: 40367530-40367530 on Build GRCh38
rs200444231 Chr.15: 40367578-40367578 on Build GRCh38
rs758069989 Chr.15: 40367568-40367568 on Build GRCh38
rs768114606 Chr.15: 40367550-40367550 on Build GRCh38
rs141024022 Chr.15: 40367532-40367532 on Build GRCh38
rs371735197 Chr.15: 40367475-40367475 on Build GRCh38
rs141482002 Chr.15: 40367387-40367387 on Build GRCh38
rs377372901 Chr.15: 40367482-40367482 on Build GRCh38
rs751050430 Chr.15: 40367574-40367574 on Build GRCh38
rs35004278 Chr.15: 40367512-40367512 on Build GRCh38
rs769887184 Chr.15: 40367469-40367469 on Build GRCh38
rs142190316 Chr.15: 40367519-40367519 on Build GRCh38
rs375825313 Chr.15: 40367587-40367587 on Build GRCh38
rs757793651 Chr.15: 40367508-40367508 on Build GRCh38
rs753473635 Chr.15: 40367375-40367375 on Build GRCh38

View Full Product Details