Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46010-46259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110C
Entrez Gene ID
642273
Gene Name
family with sequence similarity 110 member C
Gene Aliases
-
UniGene
Hs.8379
Gene Chromosome Location
Chr.2: 38814-47077 on Build GRCh38
Transcripts
NM_001077710.2 XM_011510372.2 XM_017004689.1 XM_017004690.1 XM_017004691.1 XM_017004692.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5476384 c.365T>C p.L122P Chr.2: 46021-46021 on Build GRCh38
5615096 c.346G>T p.D116Y Chr.2: 46040-46040 on Build GRCh38
1306826 c.297G>A p.S99S Chr.2: 46089-46089 on Build GRCh38
1133006 c.307T>C p.Y103H Chr.2: 46079-46079 on Build GRCh38
5354456 c.196G>A p.G66R Chr.2: 46190-46190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756787560 Chr.2: 46143-46143 on Build GRCh38
rs774423912 Chr.2: 46104-46104 on Build GRCh38
rs555134221 Chr.2: 46238-46238 on Build GRCh38
rs761925817 Chr.2: 46098-46098 on Build GRCh38
rs752543996 Chr.2: 46121-46121 on Build GRCh38
rs758912439 Chr.2: 46150-46150 on Build GRCh38
rs756767817 Chr.2: 46065-46065 on Build GRCh38
rs748381670 Chr.2: 46160-46160 on Build GRCh38
rs752096141 Chr.2: 46235-46235 on Build GRCh38
rs372922437 Chr.2: 46075-46075 on Build GRCh38
rs986801248 Chr.2: 46241-46241 on Build GRCh38
rs759603340 Chr.2: 46105-46105 on Build GRCh38
rs760488460 Chr.2: 46199-46199 on Build GRCh38
rs369284985 Chr.2: 46044-46044 on Build GRCh38
rs775376908 Chr.2: 46192-46192 on Build GRCh38
rs747399609 Chr.2: 46156-46156 on Build GRCh38
rs202148051 Chr.2: 46145-46145 on Build GRCh38
rs758857104 Chr.2: 46228-46228 on Build GRCh38
rs777080652 Chr.2: 46055-46055 on Build GRCh38
rs780360256 Chr.2: 46152-46152 on Build GRCh38
rs371584574 Chr.2: 46215-46215 on Build GRCh38
rs768563535 Chr.2: 46036-46036 on Build GRCh38
rs752382726 Chr.2: 46054-46054 on Build GRCh38
rs376180184 Chr.2: 46079-46079 on Build GRCh38
rs968265706 Chr.2: 46168-46168 on Build GRCh38
rs776334874 Chr.2: 46205-46205 on Build GRCh38
rs769801135 Chr.2: 46103-46103 on Build GRCh38
rs745732030 Chr.2: 46185-46185 on Build GRCh38
rs759370474 Chr.2: 46048-46048 on Build GRCh38
rs763862426 Chr.2: 46201-46201 on Build GRCh38
rs768955675 Chr.2: 46158-46158 on Build GRCh38
rs749969357 Chr.2: 46221-46221 on Build GRCh38
rs773436812 Chr.2: 46175-46175 on Build GRCh38
rs772166123 Chr.2: 46190-46190 on Build GRCh38
rs975453484 Chr.2: 46170-46170 on Build GRCh38
rs377079549 Chr.2: 46037-46037 on Build GRCh38
rs915843500 Chr.2: 46223-46223 on Build GRCh38
rs781440688 Chr.2: 46159-46159 on Build GRCh38
rs957138980 Chr.2: 46135-46135 on Build GRCh38
rs747170500 Chr.2: 46090-46090 on Build GRCh38
rs776528509 Chr.2: 46096-46096 on Build GRCh38
rs988420460 Chr.2: 46140-46140 on Build GRCh38
rs761388273 Chr.2: 46218-46218 on Build GRCh38
rs753419882 Chr.2: 46062-46062 on Build GRCh38
rs769900889 Chr.2: 46171-46171 on Build GRCh38
rs753474404 Chr.2: 46142-46142 on Build GRCh38
rs565721757 Chr.2: 46219-46219 on Build GRCh38
rs768957501 Chr.2: 46092-46092 on Build GRCh38
rs767482919 Chr.2: 46052-46052 on Build GRCh38
rs956096350 Chr.2: 46236-46236 on Build GRCh38
rs772415256 Chr.2: 46077-46077 on Build GRCh38
rs536007489 Chr.2: 46230-46230 on Build GRCh38
rs760362480 Chr.2: 46129-46129 on Build GRCh38
rs766365387 Chr.2: 46043-46043 on Build GRCh38
rs777612092 Chr.2: 46198-46199 on Build GRCh38
rs763828716 Chr.2: 46141-46141 on Build GRCh38
rs751496838 Chr.2: 46047-46047 on Build GRCh38
rs957289912 Chr.2: 46191-46191 on Build GRCh38
rs762763448 Chr.2: 46038-46038 on Build GRCh38
rs370633624 Chr.2: 46120-46120 on Build GRCh38

View Full Product Details