Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66564074-66564303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
UniGene
Hs.11590
Gene Chromosome Location
Chr.11: 66563463-66568576 on Build GRCh38
Transcripts
NM_003793.3 XM_011545328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475787 c.1360G>A p.G454S Chr.11: 66564108-66564108 on Build GRCh38
3716852 c.1330G>A p.V444I Chr.11: 66564138-66564138 on Build GRCh38
7221177 c.1329C>T p.D443D Chr.11: 66564139-66564139 on Build GRCh38
358562 c.1379A>T p.K460M Chr.11: 66564089-66564089 on Build GRCh38
3953689 c.1366G>A p.D456N Chr.11: 66564102-66564102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770836708 Chr.11: 66564146-66564146 on Build GRCh38
rs148155987 Chr.11: 66564100-66564100 on Build GRCh38
rs73505406 Chr.11: 66564165-66564165 on Build GRCh38
rs397514732 Chr.11: 66564095-66564095 on Build GRCh38
rs189448201 Chr.11: 66564284-66564284 on Build GRCh38
rs988896336 Chr.11: 66564200-66564200 on Build GRCh38
rs951973967 Chr.11: 66564266-66564266 on Build GRCh38
rs142523550 Chr.11: 66564134-66564134 on Build GRCh38
rs759434213 Chr.11: 66564160-66564160 on Build GRCh38
rs774367919 Chr.11: 66564138-66564138 on Build GRCh38
rs183930092 Chr.11: 66564278-66564278 on Build GRCh38
rs111799708 Chr.11: 66564241-66564241 on Build GRCh38
rs373410131 Chr.11: 66564175-66564175 on Build GRCh38
rs749483471 Chr.11: 66564130-66564130 on Build GRCh38
rs1033685612 Chr.11: 66564170-66564170 on Build GRCh38
rs17147739 Chr.11: 66564251-66564251 on Build GRCh38
rs758616720 Chr.11: 66564142-66564143 on Build GRCh38
rs1013610775 Chr.11: 66564171-66564171 on Build GRCh38
rs888414145 Chr.11: 66564098-66564098 on Build GRCh38
rs1023954735 Chr.11: 66564214-66564214 on Build GRCh38
rs927094585 Chr.11: 66564265-66564265 on Build GRCh38
rs776303065 Chr.11: 66564151-66564151 on Build GRCh38
rs377538892 Chr.11: 66564176-66564176 on Build GRCh38
rs746980424 Chr.11: 66564139-66564139 on Build GRCh38
rs780392718 Chr.11: 66564119-66564119 on Build GRCh38
rs940645860 Chr.11: 66564279-66564279 on Build GRCh38
rs11227533 Chr.11: 66564195-66564195 on Build GRCh38
rs772207393 Chr.11: 66564166-66564166 on Build GRCh38
rs34940268 Chr.11: 66564270-66564270 on Build GRCh38
rs984701755 Chr.11: 66564275-66564275 on Build GRCh38

View Full Product Details