Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34076095-34076346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN1
Entrez Gene ID
4076
Gene Name
cell cycle associated protein 1
Gene Aliases
GPIAP1, GPIP137, GRIP137, M11S1, RNG105, p137GPI
UniGene
Hs.471818
Gene Chromosome Location
Chr.11: 34051617-34102610 on Build GRCh38
Transcripts
NM_005898.4 NM_203364.2 XM_017017749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2115414 c.456A>G p.L152L Chr.11: 34076325-34076325 on Build GRCh38
926583 c.448C>T p.L150F Chr.11: 34076317-34076317 on Build GRCh38
1152351 c.448C>T p.L150F Chr.11: 34076317-34076317 on Build GRCh38
7107733 c.451G>A p.E151K Chr.11: 34076320-34076320 on Build GRCh38
4848653 c.370C>G p.Q124E Chr.11: 34076239-34076239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762114750 Chr.11: 34076250-34076250 on Build GRCh38
rs773560298 Chr.11: 34076288-34076288 on Build GRCh38
rs531864201 Chr.11: 34076166-34076166 on Build GRCh38
rs533082069 Chr.11: 34076226-34076226 on Build GRCh38
rs774173477 Chr.11: 34076216-34076216 on Build GRCh38
rs746394008 Chr.11: 34076171-34076171 on Build GRCh38
rs756724979 Chr.11: 34076129-34076129 on Build GRCh38
rs374713036 Chr.11: 34076182-34076182 on Build GRCh38
rs199988323 Chr.11: 34076303-34076303 on Build GRCh38
rs763258923 Chr.11: 34076293-34076293 on Build GRCh38
rs780640409 Chr.11: 34076145-34076145 on Build GRCh38
rs201634114 Chr.11: 34076261-34076261 on Build GRCh38
rs898619029 Chr.11: 34076155-34076155 on Build GRCh38
rs759571453 Chr.11: 34076222-34076222 on Build GRCh38
rs770857389 Chr.11: 34076215-34076215 on Build GRCh38
rs776921720 Chr.11: 34076248-34076248 on Build GRCh38
rs569568544 Chr.11: 34076193-34076194 on Build GRCh38
rs779213267 Chr.11: 34076192-34076193 on Build GRCh38
rs1047532741 Chr.11: 34076217-34076217 on Build GRCh38
rs954676326 Chr.11: 34076194-34076194 on Build GRCh38
rs769492425 Chr.11: 34076197-34076197 on Build GRCh38
rs747793733 Chr.11: 34076196-34076196 on Build GRCh38
rs772268565 Chr.11: 34076313-34076314 on Build GRCh38
rs768827790 Chr.11: 34076224-34076224 on Build GRCh38
rs367546708 Chr.11: 34076209-34076209 on Build GRCh38
rs765572855 Chr.11: 34076274-34076274 on Build GRCh38
rs996046166 Chr.11: 34076138-34076138 on Build GRCh38
rs780720143 Chr.11: 34076189-34076189 on Build GRCh38
rs746612606 Chr.11: 34076126-34076126 on Build GRCh38
rs777484454 Chr.11: 34076207-34076207 on Build GRCh38
rs751978609 Chr.11: 34076313-34076313 on Build GRCh38
rs904388544 Chr.11: 34076127-34076127 on Build GRCh38

View Full Product Details