Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171841596-171841855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
UniGene
Hs.654775
Gene Chromosome Location
Chr.1: 171841478-172418466 on Build GRCh38
Transcripts
NM_001136127.2 NM_001278252.1 NM_015569.4 XM_005245079.1 XM_005245080.1 XM_006711268.2 XM_017000976.1 XM_017000977.1 XM_017000978.1 XM_017000979.1 XM_017000981.1 XM_017000982.1 XM_017000983.1 XM_017000984.1 XM_017000985.1 XM_017000988.1 XM_017000989.1 XM_017000990.1 XM_017000991.1 XM_017000992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1336254 c.32C>A p.P11Q Chr.1: 171841688-171841688 on Build GRCh38
382984 c.161G>T p.R54M Chr.1: 171841817-171841817 on Build GRCh38
4632579 c.110T>A p.V37E Chr.1: 171841766-171841766 on Build GRCh38
6541248 c.57G>T p.A19A Chr.1: 171841713-171841713 on Build GRCh38
6647656 c.66G>A p.A22A Chr.1: 171841722-171841722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759878201 Chr.1: 171841625-171841625 on Build GRCh38
rs943246549 Chr.1: 171841794-171841794 on Build GRCh38
rs747917880 Chr.1: 171841686-171841686 on Build GRCh38
rs778559685 Chr.1: 171841800-171841800 on Build GRCh38
rs558450227 Chr.1: 171841830-171841830 on Build GRCh38
rs369144398 Chr.1: 171841699-171841699 on Build GRCh38
rs376123360 Chr.1: 171841689-171841689 on Build GRCh38
rs753168437 Chr.1: 171841632-171841632 on Build GRCh38
rs200086525 Chr.1: 171841835-171841835 on Build GRCh38
rs776921771 Chr.1: 171841703-171841703 on Build GRCh38
rs771052290 Chr.1: 171841825-171841825 on Build GRCh38
rs758190686 Chr.1: 171841821-171841821 on Build GRCh38
rs754725721 Chr.1: 171841667-171841667 on Build GRCh38
rs764385995 Chr.1: 171841728-171841728 on Build GRCh38
rs754458688 Chr.1: 171841780-171841780 on Build GRCh38
rs373357481 Chr.1: 171841823-171841823 on Build GRCh38
rs375449968 Chr.1: 171841761-171841761 on Build GRCh38
rs569863675 Chr.1: 171841785-171841785 on Build GRCh38
rs748904460 Chr.1: 171841666-171841666 on Build GRCh38
rs200969795 Chr.1: 171841764-171841764 on Build GRCh38
rs752034781 Chr.1: 171841739-171841739 on Build GRCh38
rs754091369 Chr.1: 171841618-171841618 on Build GRCh38
rs373374370 Chr.1: 171841722-171841722 on Build GRCh38
rs371816248 Chr.1: 171841639-171841639 on Build GRCh38
rs958126966 Chr.1: 171841719-171841719 on Build GRCh38
rs755570585 Chr.1: 171841649-171841649 on Build GRCh38
rs376508475 Chr.1: 171841826-171841826 on Build GRCh38
rs771766621 Chr.1: 171841687-171841687 on Build GRCh38
rs12239702 Chr.1: 171841820-171841820 on Build GRCh38
rs775729513 Chr.1: 171841833-171841833 on Build GRCh38
rs966543689 Chr.1: 171841773-171841773 on Build GRCh38
rs758705392 Chr.1: 171841633-171841633 on Build GRCh38
rs759823278 Chr.1: 171841713-171841713 on Build GRCh38
rs778559191 Chr.1: 171841681-171841681 on Build GRCh38
rs771104962 Chr.1: 171841700-171841700 on Build GRCh38
rs1020763141 Chr.1: 171841712-171841712 on Build GRCh38
rs958082127 Chr.1: 171841680-171841680 on Build GRCh38
rs763604485 Chr.1: 171841832-171841832 on Build GRCh38
rs757798537 Chr.1: 171841752-171841752 on Build GRCh38
rs765444702 Chr.1: 171841718-171841718 on Build GRCh38
rs774761654 Chr.1: 171841836-171841836 on Build GRCh38
rs752089668 Chr.1: 171841641-171841641 on Build GRCh38
rs765519410 Chr.1: 171841626-171841626 on Build GRCh38
rs773119526 Chr.1: 171841812-171841812 on Build GRCh38

View Full Product Details