Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38074288-38074561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1B1
Entrez Gene ID
1545
Gene Name
cytochrome P450 family 1 subfamily B member 1
Gene Aliases
CP1B, CYPIB1, GLC3A, P4501B1
UniGene
Hs.154654
Gene Chromosome Location
Chr.2: 38067603-38076181 on Build GRCh38
Transcripts
NM_000104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
293627 c.868_869insC p.R290fs*37 Chr.2: 38074520-38074521 on Build GRCh38
1249566 c.870C>T p.R290R Chr.2: 38074519-38074519 on Build GRCh38
6639050 c.981C>A p.I327I Chr.2: 38074408-38074408 on Build GRCh38
4583492 c.873C>T p.D291D Chr.2: 38074516-38074516 on Build GRCh38
4562231 c.917G>A p.G306E Chr.2: 38074472-38074472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748925506 Chr.2: 38074346-38074346 on Build GRCh38
rs753629193 Chr.2: 38074460-38074460 on Build GRCh38
rs778482216 Chr.2: 38074357-38074357 on Build GRCh38
rs1037865580 Chr.2: 38074486-38074486 on Build GRCh38
rs760704189 Chr.2: 38074513-38074513 on Build GRCh38
rs764907266 Chr.2: 38074464-38074464 on Build GRCh38
rs967256395 Chr.2: 38074375-38074375 on Build GRCh38
rs777678299 Chr.2: 38074404-38074404 on Build GRCh38
rs368249322 Chr.2: 38074539-38074539 on Build GRCh38
rs775607969 Chr.2: 38074453-38074453 on Build GRCh38
rs747529058 Chr.2: 38074360-38074360 on Build GRCh38
rs762129923 Chr.2: 38074370-38074370 on Build GRCh38
rs762178467 Chr.2: 38074340-38074344 on Build GRCh38
rs749521942 Chr.2: 38074442-38074442 on Build GRCh38
rs760687362 Chr.2: 38074454-38074454 on Build GRCh38
rs112059845 Chr.2: 38074445-38074445 on Build GRCh38
rs72480439 Chr.2: 38074517-38074517 on Build GRCh38
rs754302280 Chr.2: 38074422-38074422 on Build GRCh38
rs748916597 Chr.2: 38074526-38074526 on Build GRCh38
rs371549103 Chr.2: 38074393-38074393 on Build GRCh38
rs764056142 Chr.2: 38074515-38074515 on Build GRCh38
rs777516833 Chr.2: 38074323-38074323 on Build GRCh38
rs775700013 Chr.2: 38074512-38074512 on Build GRCh38
rs750132418 Chr.2: 38074533-38074533 on Build GRCh38
rs771079700 Chr.2: 38074484-38074484 on Build GRCh38
rs556393307 Chr.2: 38074426-38074426 on Build GRCh38
rs773550695 Chr.2: 38074441-38074441 on Build GRCh38
rs587778875 Chr.2: 38074520-38074521 on Build GRCh38
rs750936934 Chr.2: 38074480-38074482 on Build GRCh38
rs375173625 Chr.2: 38074522-38074522 on Build GRCh38
rs753612739 Chr.2: 38074411-38074411 on Build GRCh38
rs764041395 Chr.2: 38074458-38074458 on Build GRCh38
rs774324646 Chr.2: 38074448-38074448 on Build GRCh38
rs779403682 Chr.2: 38074469-38074469 on Build GRCh38
rs756020429 Chr.2: 38074402-38074402 on Build GRCh38
rs746045046 Chr.2: 38074498-38074498 on Build GRCh38
rs367713104 Chr.2: 38074374-38074374 on Build GRCh38
rs778110979 Chr.2: 38074477-38074477 on Build GRCh38
rs773475208 Chr.2: 38074376-38074376 on Build GRCh38
rs749928652 Chr.2: 38074465-38074465 on Build GRCh38
rs201544164 Chr.2: 38074530-38074530 on Build GRCh38
rs751000368 Chr.2: 38074543-38074543 on Build GRCh38
rs752456881 Chr.2: 38074401-38074401 on Build GRCh38
rs199836011 Chr.2: 38074438-38074438 on Build GRCh38
rs765666893 Chr.2: 38074417-38074418 on Build GRCh38
rs370935810 Chr.2: 38074490-38074490 on Build GRCh38
rs778299704 Chr.2: 38074427-38074427 on Build GRCh38
rs751595272 Chr.2: 38074382-38074382 on Build GRCh38
rs373339128 Chr.2: 38074368-38074368 on Build GRCh38
rs950785015 Chr.2: 38074457-38074457 on Build GRCh38
rs368794893 Chr.2: 38074451-38074451 on Build GRCh38
rs748708268 Chr.2: 38074471-38074471 on Build GRCh38
rs765884873 Chr.2: 38074542-38074542 on Build GRCh38
rs769149503 Chr.2: 38074361-38074361 on Build GRCh38
rs763154093 Chr.2: 38074377-38074377 on Build GRCh38
rs377539363 Chr.2: 38074478-38074478 on Build GRCh38
rs776880384 Chr.2: 38074318-38074318 on Build GRCh38
rs759392224 Chr.2: 38074384-38074384 on Build GRCh38
rs772259541 Chr.2: 38074506-38074506 on Build GRCh38
rs745318611 Chr.2: 38074430-38074430 on Build GRCh38
rs756975982 Chr.2: 38074463-38074463 on Build GRCh38
rs927496920 Chr.2: 38074536-38074536 on Build GRCh38
rs66583685 Chr.2: 38074356-38074356 on Build GRCh38
rs766501128 Chr.2: 38074381-38074381 on Build GRCh38
rs72549382 Chr.2: 38074431-38074431 on Build GRCh38
rs372321135 Chr.2: 38074472-38074472 on Build GRCh38
rs903800311 Chr.2: 38074363-38074363 on Build GRCh38
rs748634651 Chr.2: 38074433-38074433 on Build GRCh38
rs142854274 Chr.2: 38074525-38074525 on Build GRCh38
rs949383006 Chr.2: 38074468-38074468 on Build GRCh38
rs954811191 Chr.2: 38074339-38074339 on Build GRCh38
rs755861122 Chr.2: 38074311-38074311 on Build GRCh38
rs777582869 Chr.2: 38074367-38074367 on Build GRCh38

View Full Product Details