Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46484700-46484925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
UniGene
Hs.189585
Gene Chromosome Location
Chr.21: 46458942-46570900 on Build GRCh38
Transcripts
NM_001146115.1 NM_001146116.1 NM_015151.3 NM_206889.2 NM_206890.2 NM_206891.2 XM_011529492.2 XM_011529501.1 XM_011529502.1 XM_017028291.1 XM_017028292.1 XM_017028293.1 XM_017028294.1 XM_017028295.1 XM_017028296.1 XM_017028297.1 XM_017028298.1 XM_017028299.1 XM_017028300.1 XM_017028301.1 XM_017028302.1 XM_017028303.1 XM_017028304.1 XM_017028305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6328294 c.126G>A p.R42R Chr.21: 46484791-46484791 on Build GRCh38
260423 c.95A>G p.D32G Chr.21: 46484760-46484760 on Build GRCh38
6328295 c.126G>A p.R42R Chr.21: 46484791-46484791 on Build GRCh38
6094794 c.148A>C p.I50L Chr.21: 46484813-46484813 on Build GRCh38
444645 c.160C>G p.Q54E Chr.21: 46484825-46484825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559744667 Chr.21: 46484881-46484881 on Build GRCh38
rs771981002 Chr.21: 46484744-46484744 on Build GRCh38
rs753090145 Chr.21: 46484864-46484864 on Build GRCh38
rs999524196 Chr.21: 46484777-46484777 on Build GRCh38
rs1033009033 Chr.21: 46484781-46484781 on Build GRCh38
rs865966905 Chr.21: 46484789-46484789 on Build GRCh38
rs753592360 Chr.21: 46484855-46484855 on Build GRCh38
rs966711088 Chr.21: 46484767-46484767 on Build GRCh38
rs970868853 Chr.21: 46484824-46484824 on Build GRCh38
rs375195048 Chr.21: 46484817-46484817 on Build GRCh38
rs913646236 Chr.21: 46484827-46484827 on Build GRCh38
rs768459539 Chr.21: 46484834-46484834 on Build GRCh38
rs747114647 Chr.21: 46484872-46484872 on Build GRCh38
rs944517956 Chr.21: 46484784-46484784 on Build GRCh38
rs777923923 Chr.21: 46484761-46484761 on Build GRCh38
rs747118563 Chr.21: 46484793-46484793 on Build GRCh38
rs562300500 Chr.21: 46484818-46484818 on Build GRCh38
rs774228758 Chr.21: 46484837-46484837 on Build GRCh38
rs754094478 Chr.21: 46484848-46484848 on Build GRCh38
rs760168589 Chr.21: 46484847-46484847 on Build GRCh38
rs761812959 Chr.21: 46484838-46484838 on Build GRCh38
rs771182414 Chr.21: 46484808-46484808 on Build GRCh38
rs756236572 Chr.21: 46484811-46484811 on Build GRCh38
rs370847080 Chr.21: 46484735-46484735 on Build GRCh38
rs755355477 Chr.21: 46484858-46484858 on Build GRCh38
rs749934152 Chr.21: 46484843-46484843 on Build GRCh38
rs376130886 Chr.21: 46484865-46484865 on Build GRCh38
rs200756742 Chr.21: 46484860-46484860 on Build GRCh38
rs766876847 Chr.21: 46484840-46484840 on Build GRCh38
rs534623367 Chr.21: 46484879-46484880 on Build GRCh38

View Full Product Details