Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159880495-159880762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP45
Entrez Gene ID
25790
Gene Name
cilia and flagella associated protein 45
Gene Aliases
CCDC19, NESG1
UniGene
Hs.647705
Gene Chromosome Location
Chr.1: 159872364-159900116 on Build GRCh38
Transcripts
NM_012337.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3476631 c.1032C>A p.T344T Chr.1: 159880566-159880566 on Build GRCh38
3750498 c.987G>C p.L329L Chr.1: 159880611-159880611 on Build GRCh38
1688920 c.908G>A p.R303Q Chr.1: 159880690-159880690 on Build GRCh38
7359915 c.980C>A p.A327E Chr.1: 159880618-159880618 on Build GRCh38
4763202 c.978A>T p.K326N Chr.1: 159880620-159880620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143489154 Chr.1: 159880666-159880666 on Build GRCh38
rs910471941 Chr.1: 159880721-159880721 on Build GRCh38
rs758851608 Chr.1: 159880634-159880634 on Build GRCh38
rs759377667 Chr.1: 159880696-159880696 on Build GRCh38
rs374230806 Chr.1: 159880643-159880643 on Build GRCh38
rs772112494 Chr.1: 159880700-159880700 on Build GRCh38
rs774296362 Chr.1: 159880582-159880582 on Build GRCh38
rs760240240 Chr.1: 159880623-159880623 on Build GRCh38
rs7550537 Chr.1: 159880611-159880611 on Build GRCh38
rs777985657 Chr.1: 159880675-159880675 on Build GRCh38
rs139464711 Chr.1: 159880646-159880646 on Build GRCh38
rs763843109 Chr.1: 159880625-159880625 on Build GRCh38
rs749034897 Chr.1: 159880684-159880684 on Build GRCh38
rs201144590 Chr.1: 159880691-159880691 on Build GRCh38
rs777898681 Chr.1: 159880733-159880734 on Build GRCh38
rs945554552 Chr.1: 159880642-159880642 on Build GRCh38
rs770746064 Chr.1: 159880690-159880690 on Build GRCh38
rs367620563 Chr.1: 159880580-159880580 on Build GRCh38
rs753571867 Chr.1: 159880723-159880723 on Build GRCh38
rs774341505 Chr.1: 159880632-159880632 on Build GRCh38
rs943358217 Chr.1: 159880722-159880722 on Build GRCh38
rs750755248 Chr.1: 159880631-159880631 on Build GRCh38
rs766913423 Chr.1: 159880605-159880605 on Build GRCh38
rs368911610 Chr.1: 159880650-159880650 on Build GRCh38
rs774795743 Chr.1: 159880710-159880710 on Build GRCh38
rs748331211 Chr.1: 159880664-159880664 on Build GRCh38
rs138411937 Chr.1: 159880648-159880648 on Build GRCh38
rs760274621 Chr.1: 159880712-159880712 on Build GRCh38
rs144418075 Chr.1: 159880649-159880649 on Build GRCh38
rs145119988 Chr.1: 159880585-159880585 on Build GRCh38

View Full Product Details