Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 50285314-50285555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXXC1
Entrez Gene ID
30827
Gene Name
CXXC finger protein 1
Gene Aliases
2410002I16Rik, 5830420C16Rik, CFP1, CGBP, HsT2645, PCCX1, PHF18, SPP1, ZCGPC1, hCGBP
UniGene
Hs.180933
Gene Chromosome Location
Chr.18: 50282343-50288322 on Build GRCh38
Transcripts
NM_001101654.1 NM_014593.3 XM_011525940.2 XM_011525941.2 XM_017025718.1 XM_017025719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7132922 c.665C>T p.S222L Chr.18: 50285326-50285326 on Build GRCh38
3526241 c.657C>T p.F219F Chr.18: 50285334-50285334 on Build GRCh38
5158008 c.640-7G>A p.? Chr.18: 50285358-50285358 on Build GRCh38
74300 c.657C>G p.F219L Chr.18: 50285334-50285334 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs921890548 Chr.18: 50285470-50285470 on Build GRCh38
rs373953892 Chr.18: 50285398-50285398 on Build GRCh38
rs911482511 Chr.18: 50285501-50285501 on Build GRCh38
rs550879113 Chr.18: 50285418-50285418 on Build GRCh38
rs574717447 Chr.18: 50285530-50285530 on Build GRCh38
rs115234383 Chr.18: 50285519-50285519 on Build GRCh38
rs754415088 Chr.18: 50285382-50285382 on Build GRCh38
rs575692219 Chr.18: 50285500-50285501 on Build GRCh38
rs956761226 Chr.18: 50285428-50285428 on Build GRCh38
rs569935145 Chr.18: 50285444-50285444 on Build GRCh38
rs1012964259 Chr.18: 50285437-50285437 on Build GRCh38
rs552388000 Chr.18: 50285497-50285497 on Build GRCh38
rs760137943 Chr.18: 50285382-50285382 on Build GRCh38
rs772731321 Chr.18: 50285379-50285379 on Build GRCh38
rs766269244 Chr.18: 50285393-50285393 on Build GRCh38
rs914353103 Chr.18: 50285458-50285458 on Build GRCh38
rs1016074884 Chr.18: 50285408-50285408 on Build GRCh38
rs771904271 Chr.18: 50285364-50285364 on Build GRCh38
rs539064966 Chr.18: 50285487-50285487 on Build GRCh38
rs369879084 Chr.18: 50285394-50285394 on Build GRCh38
rs769129941 Chr.18: 50285358-50285358 on Build GRCh38
rs765008880 Chr.18: 50285400-50285400 on Build GRCh38
rs759409678 Chr.18: 50285431-50285431 on Build GRCh38
rs1051365417 Chr.18: 50285503-50285503 on Build GRCh38
rs2276364 Chr.18: 50285505-50285505 on Build GRCh38
rs375977156 Chr.18: 50285359-50285359 on Build GRCh38
rs962167487 Chr.18: 50285423-50285423 on Build GRCh38
rs534804373 Chr.18: 50285517-50285517 on Build GRCh38
rs947251344 Chr.18: 50285466-50285466 on Build GRCh38
rs975039031 Chr.18: 50285469-50285469 on Build GRCh38
rs1037457348 Chr.18: 50285536-50285536 on Build GRCh38
rs1022967876 Chr.18: 50285440-50285440 on Build GRCh38
rs958616923 Chr.18: 50285392-50285392 on Build GRCh38
rs112730220 Chr.18: 50285353-50285353 on Build GRCh38
rs752527434 Chr.18: 50285401-50285401 on Build GRCh38
rs778058292 Chr.18: 50285395-50285395 on Build GRCh38

View Full Product Details