Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158741482-158741749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT2
Entrez Gene ID
57488
Gene Name
extended synaptotagmin 2
Gene Aliases
CHR2SYT, E-Syt2, FAM62B
UniGene
Hs.490795
Gene Chromosome Location
Chr.7: 158730998-158829628 on Build GRCh38
Transcripts
NM_020728.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5530630 c.451C>T p.P151S Chr.7: 158741626-158741626 on Build GRCh38
6109399 c.2209G>T p.A737S Chr.7: 158741563-158741563 on Build GRCh38
7366672 c.391G>A p.E131K Chr.7: 158741686-158741686 on Build GRCh38
5407106 c.2184C>T p.I728I Chr.7: 158741588-158741588 on Build GRCh38
7135890 c.465A>T p.S155S Chr.7: 158741612-158741612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59980573 Chr.7: 158741690-158741690 on Build GRCh38
rs371929215 Chr.7: 158741566-158741566 on Build GRCh38
rs774765568 Chr.7: 158741692-158741692 on Build GRCh38
rs756105628 Chr.7: 158741646-158741646 on Build GRCh38
rs943714755 Chr.7: 158741570-158741570 on Build GRCh38
rs747377147 Chr.7: 158741588-158741588 on Build GRCh38
rs371306927 Chr.7: 158741675-158741675 on Build GRCh38
rs368676835 Chr.7: 158741663-158741663 on Build GRCh38
rs374453770 Chr.7: 158741659-158741659 on Build GRCh38
rs759600331 Chr.7: 158741518-158741518 on Build GRCh38
rs1012524437 Chr.7: 158741667-158741667 on Build GRCh38
rs138969546 Chr.7: 158741684-158741684 on Build GRCh38
rs781644750 Chr.7: 158741676-158741676 on Build GRCh38
rs773507830 Chr.7: 158741587-158741587 on Build GRCh38
rs368464151 Chr.7: 158741564-158741564 on Build GRCh38
rs779723163 Chr.7: 158741573-158741573 on Build GRCh38
rs200023069 Chr.7: 158741582-158741582 on Build GRCh38
rs748896101 Chr.7: 158741579-158741579 on Build GRCh38
rs1008310519 Chr.7: 158741632-158741632 on Build GRCh38
rs775734975 Chr.7: 158741618-158741618 on Build GRCh38
rs753010070 Chr.7: 158741526-158741526 on Build GRCh38
rs1036665528 Chr.7: 158741662-158741662 on Build GRCh38
rs779486617 Chr.7: 158741648-158741648 on Build GRCh38
rs750250873 Chr.7: 158741726-158741726 on Build GRCh38
rs750365571 Chr.7: 158741637-158741637 on Build GRCh38
rs758726644 Chr.7: 158741529-158741529 on Build GRCh38
rs763933304 Chr.7: 158741548-158741548 on Build GRCh38
rs368218045 Chr.7: 158741604-158741604 on Build GRCh38
rs767469150 Chr.7: 158741628-158741628 on Build GRCh38
rs757060506 Chr.7: 158741625-158741625 on Build GRCh38
rs771247172 Chr.7: 158741674-158741674 on Build GRCh38
rs765550564 Chr.7: 158741714-158741714 on Build GRCh38
rs764419846 Chr.7: 158741619-158741619 on Build GRCh38
rs771464234 Chr.7: 158741600-158741600 on Build GRCh38
rs890689403 Chr.7: 158741590-158741590 on Build GRCh38
rs766808427 Chr.7: 158741502-158741502 on Build GRCh38
rs757155824 Chr.7: 158741562-158741562 on Build GRCh38
rs748806082 Chr.7: 158741655-158741655 on Build GRCh38
rs77532359 Chr.7: 158741525-158741525 on Build GRCh38
rs765193309 Chr.7: 158741611-158741611 on Build GRCh38
rs780629818 Chr.7: 158741691-158741691 on Build GRCh38
rs777060478 Chr.7: 158741601-158741601 on Build GRCh38
rs374236194 Chr.7: 158741506-158741506 on Build GRCh38
rs778370657 Chr.7: 158741515-158741518 on Build GRCh38
rs57686648 Chr.7: 158741681-158741681 on Build GRCh38
rs7792976 Chr.7: 158741680-158741680 on Build GRCh38
rs781060682 Chr.7: 158741563-158741563 on Build GRCh38
rs761635374 Chr.7: 158741715-158741715 on Build GRCh38
rs114848522 Chr.7: 158741561-158741561 on Build GRCh38
rs562066737 Chr.7: 158741624-158741624 on Build GRCh38
rs34287300 Chr.7: 158741526-158741527 on Build GRCh38

View Full Product Details