Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4247046-4247307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC94
Entrez Gene ID
55702
Gene Name
coiled-coil domain containing 94
Gene Aliases
-
UniGene
Hs.21811
Gene Chromosome Location
Chr.19: 4247114-4269088 on Build GRCh38
Transcripts
NM_018074.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7182368 c.6G>T p.S2S Chr.19: 4247152-4247152 on Build GRCh38
997162 c.21A>G p.L7L Chr.19: 4247167-4247167 on Build GRCh38
997161 c.16G>A p.V6I Chr.19: 4247162-4247162 on Build GRCh38
1283987 c.16G>C p.V6L Chr.19: 4247162-4247162 on Build GRCh38
159816 c.5C>T p.S2L Chr.19: 4247151-4247151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016217505 Chr.19: 4247250-4247252 on Build GRCh38
rs371273295 Chr.19: 4247132-4247132 on Build GRCh38
rs142023122 Chr.19: 4247109-4247109 on Build GRCh38
rs1013536367 Chr.19: 4247090-4247090 on Build GRCh38
rs768276762 Chr.19: 4247088-4247088 on Build GRCh38
rs574985629 Chr.19: 4247099-4247099 on Build GRCh38
rs373676911 Chr.19: 4247114-4247114 on Build GRCh38
rs1026395057 Chr.19: 4247118-4247118 on Build GRCh38
rs769257935 Chr.19: 4247185-4247185 on Build GRCh38
rs750704084 Chr.19: 4247113-4247113 on Build GRCh38
rs756793337 Chr.19: 4247140-4247140 on Build GRCh38
rs767201886 Chr.19: 4247215-4247215 on Build GRCh38
rs8112323 Chr.19: 4247253-4247253 on Build GRCh38
rs763333909 Chr.19: 4247214-4247214 on Build GRCh38
rs773510562 Chr.19: 4247209-4247209 on Build GRCh38
rs955811790 Chr.19: 4247243-4247243 on Build GRCh38
rs761946816 Chr.19: 4247193-4247193 on Build GRCh38
rs777127587 Chr.19: 4247134-4247134 on Build GRCh38
rs569116849 Chr.19: 4247268-4247268 on Build GRCh38
rs368991444 Chr.19: 4247247-4247247 on Build GRCh38
rs1006551045 Chr.19: 4247245-4247245 on Build GRCh38
rs748838850 Chr.19: 4247137-4247137 on Build GRCh38
rs112256607 Chr.19: 4247265-4247265 on Build GRCh38
rs377694798 Chr.19: 4247126-4247126 on Build GRCh38
rs377386714 Chr.19: 4247199-4247199 on Build GRCh38
rs886064045 Chr.19: 4247151-4247151 on Build GRCh38
rs776823351 Chr.19: 4247189-4247189 on Build GRCh38
rs752573207 Chr.19: 4247123-4247123 on Build GRCh38
rs1045731638 Chr.19: 4247070-4247070 on Build GRCh38
rs896379154 Chr.19: 4247082-4247082 on Build GRCh38
rs752585808 Chr.19: 4247217-4247217 on Build GRCh38
rs79942400 Chr.19: 4247273-4247273 on Build GRCh38
rs1054909928 Chr.19: 4247077-4247077 on Build GRCh38
rs767358402 Chr.19: 4247116-4247116 on Build GRCh38
rs1028526494 Chr.19: 4247236-4247236 on Build GRCh38
rs975560200 Chr.19: 4247205-4247205 on Build GRCh38
rs374660429 Chr.19: 4247175-4247175 on Build GRCh38
rs747461638 Chr.19: 4247184-4247184 on Build GRCh38
rs532334652 Chr.19: 4247255-4247255 on Build GRCh38
rs1054754043 Chr.19: 4247093-4247093 on Build GRCh38
rs755972816 Chr.19: 4247222-4247222 on Build GRCh38
rs981525026 Chr.19: 4247274-4247274 on Build GRCh38
rs548656576 Chr.19: 4247285-4247285 on Build GRCh38
rs765431035 Chr.19: 4247202-4247202 on Build GRCh38
rs150828443 Chr.19: 4247183-4247183 on Build GRCh38
rs34322392 Chr.19: 4247170-4247170 on Build GRCh38
rs543948001 Chr.19: 4247100-4247100 on Build GRCh38
rs778491292 Chr.19: 4247165-4247165 on Build GRCh38
rs1028581941 Chr.19: 4247223-4247223 on Build GRCh38
rs556514618 Chr.19: 4247096-4247096 on Build GRCh38

View Full Product Details