Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238104523-238104788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPNL
Entrez Gene ID
339768
Gene Name
espin-like
Gene Aliases
-
UniGene
Hs.127724
Gene Chromosome Location
Chr.2: 238100310-238133287 on Build GRCh38
Transcripts
NM_194312.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
720970 c.608T>C p.L203P Chr.2: 238104778-238104778 on Build GRCh38
1692239 c.601C>T p.R201C Chr.2: 238104771-238104771 on Build GRCh38
4092723 c.529C>A p.L177M Chr.2: 238104699-238104699 on Build GRCh38
4092724 c.575A>C p.K192T Chr.2: 238104745-238104745 on Build GRCh38
3051978 c.602G>A p.R201H Chr.2: 238104772-238104772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762448923 Chr.2: 238104671-238104671 on Build GRCh38
rs753793058 Chr.2: 238104619-238104619 on Build GRCh38
rs140751353 Chr.2: 238104684-238104684 on Build GRCh38
rs557725637 Chr.2: 238104692-238104692 on Build GRCh38
rs748179923 Chr.2: 238104654-238104654 on Build GRCh38
rs144708509 Chr.2: 238104713-238104713 on Build GRCh38
rs760477137 Chr.2: 238104607-238104607 on Build GRCh38
rs146620287 Chr.2: 238104705-238104705 on Build GRCh38
rs745587704 Chr.2: 238104742-238104742 on Build GRCh38
rs777343784 Chr.2: 238104640-238104640 on Build GRCh38
rs1034279361 Chr.2: 238104722-238104722 on Build GRCh38
rs772426606 Chr.2: 238104656-238104656 on Build GRCh38
rs978491927 Chr.2: 238104549-238104549 on Build GRCh38
rs753226637 Chr.2: 238104769-238104769 on Build GRCh38
rs751187489 Chr.2: 238104676-238104676 on Build GRCh38
rs183738193 Chr.2: 238104643-238104643 on Build GRCh38
rs968695982 Chr.2: 238104562-238104562 on Build GRCh38
rs529284987 Chr.2: 238104666-238104666 on Build GRCh38
rs116421471 Chr.2: 238104629-238104629 on Build GRCh38
rs533316796 Chr.2: 238104641-238104641 on Build GRCh38
rs371780532 Chr.2: 238104683-238104683 on Build GRCh38
rs758064223 Chr.2: 238104634-238104634 on Build GRCh38
rs978998058 Chr.2: 238104570-238104570 on Build GRCh38
rs368522053 Chr.2: 238104756-238104756 on Build GRCh38
rs371819795 Chr.2: 238104762-238104762 on Build GRCh38
rs958806234 Chr.2: 238104551-238104551 on Build GRCh38
rs780471889 Chr.2: 238104648-238104648 on Build GRCh38
rs201383533 Chr.2: 238104711-238104711 on Build GRCh38
rs34046909 Chr.2: 238104670-238104670 on Build GRCh38
rs766286580 Chr.2: 238104614-238104614 on Build GRCh38
rs368175214 Chr.2: 238104660-238104660 on Build GRCh38
rs542298602 Chr.2: 238104755-238104755 on Build GRCh38
rs1000145006 Chr.2: 238104718-238104718 on Build GRCh38
rs554995928 Chr.2: 238104597-238104597 on Build GRCh38
rs763989830 Chr.2: 238104725-238104725 on Build GRCh38
rs115807387 Chr.2: 238104650-238104650 on Build GRCh38
rs747287367 Chr.2: 238104719-238104719 on Build GRCh38
rs759009212 Chr.2: 238104621-238104621 on Build GRCh38
rs958430161 Chr.2: 238104590-238104590 on Build GRCh38
rs992663373 Chr.2: 238104733-238104733 on Build GRCh38
rs111369122 Chr.2: 238104697-238104697 on Build GRCh38
rs374744174 Chr.2: 238104616-238104616 on Build GRCh38
rs765031783 Chr.2: 238104669-238104669 on Build GRCh38
rs376099769 Chr.2: 238104626-238104626 on Build GRCh38
rs72985180 Chr.2: 238104550-238104550 on Build GRCh38
rs950075476 Chr.2: 238104580-238104580 on Build GRCh38
rs773152342 Chr.2: 238104606-238104606 on Build GRCh38
rs754101349 Chr.2: 238104691-238104691 on Build GRCh38
rs775396132 Chr.2: 238104745-238104745 on Build GRCh38
rs776502051 Chr.2: 238104738-238104738 on Build GRCh38
rs767019569 Chr.2: 238104761-238104761 on Build GRCh38
rs759752195 Chr.2: 238104557-238104557 on Build GRCh38
rs763797257 Chr.2: 238104675-238104675 on Build GRCh38
rs762495856 Chr.2: 238104750-238104750 on Build GRCh38
rs1036323734 Chr.2: 238104591-238104591 on Build GRCh38
rs779156800 Chr.2: 238104653-238104653 on Build GRCh38
rs773561707 Chr.2: 238104659-238104659 on Build GRCh38
rs991406047 Chr.2: 238104560-238104560 on Build GRCh38
rs562278268 Chr.2: 238104563-238104563 on Build GRCh38
rs542125122 Chr.2: 238104579-238104579 on Build GRCh38
rs769793223 Chr.2: 238104743-238104743 on Build GRCh38
rs776250145 Chr.2: 238104667-238104667 on Build GRCh38
rs149008836 Chr.2: 238104703-238104703 on Build GRCh38

View Full Product Details