Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52653060-52653287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERO1A
Entrez Gene ID
30001
Gene Name
endoplasmic reticulum oxidoreductase 1 alpha
Gene Aliases
ERO1-L, ERO1-L-alpha, ERO1-alpha, ERO1L, ERO1LA, Ero1alpha
UniGene
Hs.525339
Gene Chromosome Location
Chr.14: 52639915-52695931 on Build GRCh38
Transcripts
NM_014584.2 XM_011536684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
117956 c.859C>T p.R287* Chr.14: 52653265-52653265 on Build GRCh38
4051283 c.1045C>T p.H349Y Chr.14: 52653079-52653079 on Build GRCh38
4148130 c.916T>C p.F306L Chr.14: 52653208-52653208 on Build GRCh38
6044980 c.1004_1005insT p.Q336fs*3 Chr.14: 52653119-52653120 on Build GRCh38
1205652 c.931G>T p.E311* Chr.14: 52653193-52653193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753676845 Chr.14: 52653182-52653182 on Build GRCh38
rs748725771 Chr.14: 52653245-52653245 on Build GRCh38
rs749646868 Chr.14: 52653251-52653251 on Build GRCh38
rs573952507 Chr.14: 52653148-52653148 on Build GRCh38
rs145722252 Chr.14: 52653158-52653158 on Build GRCh38
rs781088934 Chr.14: 52653243-52653243 on Build GRCh38
rs773413061 Chr.14: 52653146-52653146 on Build GRCh38
rs780342547 Chr.14: 52653113-52653113 on Build GRCh38
rs760499654 Chr.14: 52653161-52653161 on Build GRCh38
rs991342298 Chr.14: 52653253-52653253 on Build GRCh38
rs746133119 Chr.14: 52653109-52653109 on Build GRCh38
rs755104744 Chr.14: 52653236-52653236 on Build GRCh38
rs777391697 Chr.14: 52653100-52653100 on Build GRCh38
rs143610986 Chr.14: 52653157-52653157 on Build GRCh38
rs748551230 Chr.14: 52653140-52653140 on Build GRCh38
rs370745677 Chr.14: 52653153-52653153 on Build GRCh38
rs771216182 Chr.14: 52653255-52653255 on Build GRCh38
rs374071417 Chr.14: 52653247-52653247 on Build GRCh38
rs369212913 Chr.14: 52653116-52653116 on Build GRCh38
rs758675597 Chr.14: 52653111-52653111 on Build GRCh38
rs758763441 Chr.14: 52653184-52653184 on Build GRCh38
rs553263538 Chr.14: 52653164-52653164 on Build GRCh38
rs781727835 Chr.14: 52653130-52653130 on Build GRCh38
rs573835773 Chr.14: 52653154-52653154 on Build GRCh38
rs778403123 Chr.14: 52653125-52653126 on Build GRCh38
rs770178309 Chr.14: 52653246-52653246 on Build GRCh38
rs770556003 Chr.14: 52653151-52653151 on Build GRCh38
rs542544751 Chr.14: 52653201-52653201 on Build GRCh38

View Full Product Details