Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157430585-157430844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTIP
Entrez Gene ID
9595
Gene Name
cytohesin 1 interacting protein
Gene Aliases
B3-1, CASP, CYBR, CYTHIP, HE, PSCDBP
UniGene
Hs.270
Gene Chromosome Location
Chr.2: 157412781-157444092 on Build GRCh38
Transcripts
NM_004288.4 XM_017005386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4085810 c.440T>G p.V147G Chr.2: 157430595-157430595 on Build GRCh38
6639615 c.383-1G>A p.? Chr.2: 157430653-157430653 on Build GRCh38
5529554 c.402C>T p.I134I Chr.2: 157430633-157430633 on Build GRCh38
1613542 c.413G>T p.S138I Chr.2: 157430622-157430622 on Build GRCh38
5584965 c.406G>A p.G136S Chr.2: 157430629-157430629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028640043 Chr.2: 157430753-157430753 on Build GRCh38
rs767253791 Chr.2: 157430611-157430611 on Build GRCh38
rs970553260 Chr.2: 157430708-157430708 on Build GRCh38
rs754313107 Chr.2: 157430658-157430658 on Build GRCh38
rs962685427 Chr.2: 157430650-157430650 on Build GRCh38
rs959930171 Chr.2: 157430652-157430652 on Build GRCh38
rs150721802 Chr.2: 157430638-157430638 on Build GRCh38
rs975396650 Chr.2: 157430712-157430712 on Build GRCh38
rs779402557 Chr.2: 157430678-157430678 on Build GRCh38
rs141221938 Chr.2: 157430789-157430789 on Build GRCh38
rs752276215 Chr.2: 157430624-157430624 on Build GRCh38
rs560816933 Chr.2: 157430697-157430697 on Build GRCh38
rs747269539 Chr.2: 157430683-157430683 on Build GRCh38
rs753800293 Chr.2: 157430687-157430690 on Build GRCh38
rs973471101 Chr.2: 157430710-157430710 on Build GRCh38
rs200014154 Chr.2: 157430686-157430686 on Build GRCh38
rs757691586 Chr.2: 157430661-157430661 on Build GRCh38
rs993053789 Chr.2: 157430681-157430681 on Build GRCh38
rs920630684 Chr.2: 157430734-157430734 on Build GRCh38

View Full Product Details