Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119298646-119298913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
-
UniGene
Hs.173705
Gene Chromosome Location
Chr.4: 119296419-119304445 on Build GRCh38
Transcripts
NM_001001701.3 NM_001170330.1

SNP Detail

SNP ID Chromosome Location
rs899106390 Chr.4: 119298852-119298852 on Build GRCh38
rs749359847 Chr.4: 119298772-119298772 on Build GRCh38
rs199817255 Chr.4: 119298756-119298756 on Build GRCh38
rs902819227 Chr.4: 119298700-119298700 on Build GRCh38
rs191015720 Chr.4: 119298807-119298807 on Build GRCh38
rs951184228 Chr.4: 119298806-119298806 on Build GRCh38
rs769058771 Chr.4: 119298840-119298840 on Build GRCh38
rs373952403 Chr.4: 119298767-119298767 on Build GRCh38
rs763680957 Chr.4: 119298763-119298763 on Build GRCh38
rs753496590 Chr.4: 119298764-119298764 on Build GRCh38
rs766835752 Chr.4: 119298749-119298749 on Build GRCh38
rs1026323898 Chr.4: 119298856-119298856 on Build GRCh38
rs763496175 Chr.4: 119298746-119298746 on Build GRCh38
rs893646608 Chr.4: 119298732-119298732 on Build GRCh38
rs774631641 Chr.4: 119298883-119298883 on Build GRCh38
rs769947780 Chr.4: 119298744-119298744 on Build GRCh38
rs530448424 Chr.4: 119298670-119298670 on Build GRCh38
rs759954440 Chr.4: 119298762-119298762 on Build GRCh38
rs769958930 Chr.4: 119298831-119298831 on Build GRCh38
rs758348098 Chr.4: 119298785-119298785 on Build GRCh38
rs186567016 Chr.4: 119298771-119298771 on Build GRCh38
rs993793866 Chr.4: 119298855-119298855 on Build GRCh38
rs748402538 Chr.4: 119298822-119298822 on Build GRCh38
rs998961698 Chr.4: 119298694-119298694 on Build GRCh38
rs1031227449 Chr.4: 119298715-119298715 on Build GRCh38
rs370809681 Chr.4: 119298685-119298685 on Build GRCh38
rs779911514 Chr.4: 119298797-119298797 on Build GRCh38
rs768574129 Chr.4: 119298809-119298809 on Build GRCh38
rs773541793 Chr.4: 119298745-119298745 on Build GRCh38
rs533564068 Chr.4: 119298811-119298811 on Build GRCh38

View Full Product Details