Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82157819-82158048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC57
Entrez Gene ID
284001
Gene Name
coiled-coil domain containing 57
Gene Aliases
-
UniGene
Hs.631724
Gene Chromosome Location
Chr.17: 82101458-82212892 on Build GRCh38
Transcripts
NM_001316321.1 NM_198082.2 XM_011523545.2 XM_011523546.2 XM_011523548.2 XM_011523549.2 XM_011523551.2 XM_011523552.2 XM_011523555.2 XM_011523556.2 XM_011523557.2 XM_017024462.1 XM_017024463.1 XM_017024464.1 XM_017024465.1 XM_017024466.1 XM_017024467.1 XM_017024468.1 XM_017024469.1 XM_017024470.1 XM_017024471.1 XM_017024472.1 XM_017024473.1 XM_017024474.1 XM_017024475.1 XM_017024476.1 XM_017024477.1 XM_017024478.1 XM_017024479.1 XM_017024483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4926606 c.2134C>T p.Q712* Chr.17: 82157855-82157855 on Build GRCh38
4632173 c.2096T>C p.V699A Chr.17: 82157893-82157893 on Build GRCh38
4632176 c.2078G>A p.R693H Chr.17: 82157911-82157911 on Build GRCh38
4926607 c.2134C>T p.Q712* Chr.17: 82157855-82157855 on Build GRCh38
4632174 c.2096T>C p.V699A Chr.17: 82157893-82157893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972494696 Chr.17: 82158020-82158020 on Build GRCh38
rs890049346 Chr.17: 82157849-82157849 on Build GRCh38
rs367918124 Chr.17: 82157865-82157865 on Build GRCh38
rs551236525 Chr.17: 82157942-82157942 on Build GRCh38
rs548988993 Chr.17: 82157922-82157922 on Build GRCh38
rs998359629 Chr.17: 82157839-82157839 on Build GRCh38
rs371490268 Chr.17: 82157879-82157879 on Build GRCh38
rs758062064 Chr.17: 82157992-82157992 on Build GRCh38
rs200005504 Chr.17: 82157888-82157888 on Build GRCh38
rs751979282 Chr.17: 82157912-82157912 on Build GRCh38
rs752702265 Chr.17: 82157899-82157899 on Build GRCh38
rs548915811 Chr.17: 82157846-82157846 on Build GRCh38
rs1052009708 Chr.17: 82157932-82157932 on Build GRCh38
rs867753825 Chr.17: 82157915-82157915 on Build GRCh38
rs772689360 Chr.17: 82157938-82157938 on Build GRCh38
rs147432414 Chr.17: 82157984-82157984 on Build GRCh38
rs538854935 Chr.17: 82157934-82157934 on Build GRCh38
rs981975539 Chr.17: 82157926-82157926 on Build GRCh38
rs546761432 Chr.17: 82157900-82157900 on Build GRCh38
rs781506740 Chr.17: 82157917-82157917 on Build GRCh38
rs759601417 Chr.17: 82157955-82157955 on Build GRCh38
rs778553453 Chr.17: 82157925-82157925 on Build GRCh38
rs747545285 Chr.17: 82157931-82157931 on Build GRCh38
rs766440295 Chr.17: 82157860-82157860 on Build GRCh38
rs922365936 Chr.17: 82158027-82158027 on Build GRCh38
rs372516791 Chr.17: 82157948-82157948 on Build GRCh38
rs369494382 Chr.17: 82157941-82157941 on Build GRCh38
rs939669604 Chr.17: 82157999-82157999 on Build GRCh38
rs373713185 Chr.17: 82157853-82157853 on Build GRCh38
rs779610549 Chr.17: 82158028-82158028 on Build GRCh38
rs938497290 Chr.17: 82157843-82157843 on Build GRCh38
rs756648717 Chr.17: 82157944-82157944 on Build GRCh38
rs955844980 Chr.17: 82158008-82158008 on Build GRCh38
rs746425037 Chr.17: 82157921-82157921 on Build GRCh38

View Full Product Details