Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109727224-109727481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIG4
Entrez Gene ID
9896
Gene Name
FIG4 phosphoinositide 5-phosphatase
Gene Aliases
ALS11, BTOP, CMT4J, KIAA0274, SAC3, YVS, dJ249I4.1
UniGene
Hs.529959
Gene Chromosome Location
Chr.6: 109691216-109825431 on Build GRCh38
Transcripts
NM_014845.5 XM_011536281.2 XM_017011591.1 XM_017011592.1 XM_017011593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1672670 c.412G>T p.D138Y Chr.6: 109727231-109727231 on Build GRCh38
3941462 c.358C>T p.R120W Chr.6: 109727240-109727240 on Build GRCh38
3941461 c.421C>T p.R141W Chr.6: 109727240-109727240 on Build GRCh38
4349469 c.349G>T p.D117Y Chr.6: 109727231-109727231 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779033720 Chr.6: 109727300-109727300 on Build GRCh38
rs796900144 Chr.6: 109727287-109727288 on Build GRCh38
rs572627513 Chr.6: 109727441-109727441 on Build GRCh38
rs370857139 Chr.6: 109727270-109727270 on Build GRCh38
rs746139833 Chr.6: 109727307-109727307 on Build GRCh38
rs567294037 Chr.6: 109727324-109727324 on Build GRCh38
rs867400590 Chr.6: 109727265-109727265 on Build GRCh38
rs770188660 Chr.6: 109727417-109727417 on Build GRCh38
rs965616904 Chr.6: 109727283-109727283 on Build GRCh38
rs1040386089 Chr.6: 109727393-109727394 on Build GRCh38
rs975523794 Chr.6: 109727346-109727346 on Build GRCh38
rs759482200 Chr.6: 109727291-109727292 on Build GRCh38
rs908190068 Chr.6: 109727432-109727432 on Build GRCh38
rs933692955 Chr.6: 109727440-109727440 on Build GRCh38
rs778693746 Chr.6: 109727370-109727370 on Build GRCh38
rs535283923 Chr.6: 109727443-109727443 on Build GRCh38
rs956770641 Chr.6: 109727365-109727372 on Build GRCh38
rs781309177 Chr.6: 109727277-109727277 on Build GRCh38
rs536319627 Chr.6: 109727377-109727377 on Build GRCh38
rs770434527 Chr.6: 109727294-109727295 on Build GRCh38
rs931025031 Chr.6: 109727396-109727396 on Build GRCh38
rs536626725 Chr.6: 109727248-109727248 on Build GRCh38
rs752613836 Chr.6: 109727278-109727278 on Build GRCh38
rs397821151 Chr.6: 109727297-109727298 on Build GRCh38
rs982094703 Chr.6: 109727320-109727320 on Build GRCh38
rs563921466 Chr.6: 109727287-109727287 on Build GRCh38
rs201642753 Chr.6: 109727268-109727268 on Build GRCh38
rs942860970 Chr.6: 109727390-109727390 on Build GRCh38
rs532856136 Chr.6: 109727287-109727288 on Build GRCh38
rs199593454 Chr.6: 109727298-109727298 on Build GRCh38
rs984316652 Chr.6: 109727408-109727408 on Build GRCh38
rs142893576 Chr.6: 109727389-109727389 on Build GRCh38
rs11459279 Chr.6: 109727286-109727287 on Build GRCh38
rs190287033 Chr.6: 109727274-109727274 on Build GRCh38
rs757422452 Chr.6: 109727284-109727284 on Build GRCh38
rs956953729 Chr.6: 109727269-109727269 on Build GRCh38

View Full Product Details