Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 216564160-216564389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRG
Entrez Gene ID
2104
Gene Name
estrogen related receptor gamma
Gene Aliases
ERR3, ERRgamma, NR3B3
UniGene
Hs.444225
Gene Chromosome Location
Chr.1: 216503246-217137757 on Build GRCh38
Transcripts
NM_001134285.2 NM_001243505.1 NM_001243506.1 NM_001243507.1 NM_001243509.1 NM_001243510.2 NM_001243511.2 NM_001243512.1 NM_001243513.1 NM_001243514.1 NM_001243515.1 NM_001243518.1 NM_001243519.1 NM_001438.3 NM_206594.2 NM_206595.2 XM_011509265.2 XM_011509266.2 XM_011509267.1 XM_011509268.2 XM_011509269.2 XM_011509270.1 XM_011509271.2 XM_011509274.1 XM_011509275.1 XM_011509276.1 XM_011509277.1 XM_011509278.1 XM_011509279.1 XM_011509280.2 XM_017000621.1 XM_017000622.1 XM_017000623.1 XM_017000624.1 XM_017000625.1 XM_017000626.1 XM_017000627.1 XM_017000628.1 XM_017000629.1 XM_017000630.1 XM_017000631.1 XM_017000632.1 XM_017000633.1 XM_017000634.1 XM_017000635.1 XM_017000636.1 XM_017000637.1 XM_017000638.1 XM_017000639.1 XM_017000640.1 XM_017000641.1 XM_017000642.1 XM_017000643.1 XM_017000644.1 XM_017000645.1 XM_017000646.1 XM_017000647.1 XM_017000648.1 XM_017000649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146771 c.790G>T p.A264S Chr.1: 216564291-216564291 on Build GRCh38
5460759 c.773C>T p.P258L Chr.1: 216564308-216564308 on Build GRCh38
7234194 c.653A>G p.D218G Chr.1: 216564380-216564380 on Build GRCh38
4511018 c.819C>T p.D273D Chr.1: 216564262-216564262 on Build GRCh38
376529 c.756G>T p.M252I Chr.1: 216564325-216564325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770733689 Chr.1: 216564300-216564300 on Build GRCh38
rs763892661 Chr.1: 216564189-216564189 on Build GRCh38
rs779064396 Chr.1: 216564256-216564256 on Build GRCh38
rs754403371 Chr.1: 216564242-216564242 on Build GRCh38
rs763664788 Chr.1: 216564344-216564344 on Build GRCh38
rs748236147 Chr.1: 216564258-216564258 on Build GRCh38
rs755377749 Chr.1: 216564244-216564244 on Build GRCh38
rs201736793 Chr.1: 216564265-216564265 on Build GRCh38
rs762615151 Chr.1: 216564343-216564343 on Build GRCh38
rs201435029 Chr.1: 216564332-216564332 on Build GRCh38
rs966328648 Chr.1: 216564310-216564310 on Build GRCh38
rs762859693 Chr.1: 216564252-216564252 on Build GRCh38
rs751248484 Chr.1: 216564211-216564211 on Build GRCh38
rs139709396 Chr.1: 216564259-216564259 on Build GRCh38
rs979981316 Chr.1: 216564313-216564313 on Build GRCh38
rs747209048 Chr.1: 216564285-216564285 on Build GRCh38
rs200261491 Chr.1: 216564306-216564306 on Build GRCh38
rs764293924 Chr.1: 216564192-216564192 on Build GRCh38
rs150595557 Chr.1: 216564307-216564307 on Build GRCh38
rs780520471 Chr.1: 216564232-216564232 on Build GRCh38
rs1036013279 Chr.1: 216564326-216564326 on Build GRCh38
rs756583718 Chr.1: 216564221-216564221 on Build GRCh38
rs970280342 Chr.1: 216564264-216564264 on Build GRCh38
rs149670038 Chr.1: 216564357-216564357 on Build GRCh38

View Full Product Details