Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10430393-10430642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APITD1
Entrez Gene ID
378708
Gene Name
apoptosis-inducing, TAF9-like domain 1
Gene Aliases
CENP-S, CENPS, FAAP16, MHF1
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10442815 on Build GRCh38
Transcripts
NM_199294.2 NM_001270517.1 NM_198544.3 NM_199006.2

Gene Symbol
APITD1-CORT
Entrez Gene ID
100526739
Gene Name
APITD1-CORT readthrough
Gene Aliases
-
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10452003 on Build GRCh38
Transcripts
NM_199294.2 NM_001270517.1 NM_198544.3 NM_199006.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
893258 c.39C>A p.F13L Chr.1: 10430556-10430556 on Build GRCh38
6361580 c.22G>A p.E8K Chr.1: 10430539-10430539 on Build GRCh38
6361581 c.22G>A p.E8K Chr.1: 10430539-10430539 on Build GRCh38
1583173 c.39C>A p.F13L Chr.1: 10430556-10430556 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748065363 Chr.1: 10430480-10430480 on Build GRCh38
rs775531844 Chr.1: 10430502-10430502 on Build GRCh38
rs758140060 Chr.1: 10430587-10430588 on Build GRCh38
rs1056204903 Chr.1: 10430470-10430470 on Build GRCh38
rs976265830 Chr.1: 10430550-10430550 on Build GRCh38
rs547690900 Chr.1: 10430598-10430598 on Build GRCh38
rs939255074 Chr.1: 10430620-10430620 on Build GRCh38
rs763120551 Chr.1: 10430569-10430569 on Build GRCh38
rs544729355 Chr.1: 10430418-10430418 on Build GRCh38
rs926562031 Chr.1: 10430610-10430610 on Build GRCh38
rs909594990 Chr.1: 10430578-10430578 on Build GRCh38
rs779042321 Chr.1: 10430475-10430475 on Build GRCh38
rs570419369 Chr.1: 10430576-10430576 on Build GRCh38
rs951145733 Chr.1: 10430556-10430556 on Build GRCh38
rs919412853 Chr.1: 10430551-10430551 on Build GRCh38
rs661256 Chr.1: 10430482-10430482 on Build GRCh38
rs1018281226 Chr.1: 10430532-10430532 on Build GRCh38
rs116395144 Chr.1: 10430465-10430465 on Build GRCh38
rs372438339 Chr.1: 10430579-10430579 on Build GRCh38
rs1046757246 Chr.1: 10430412-10430412 on Build GRCh38
rs897620492 Chr.1: 10430589-10430589 on Build GRCh38
rs772057575 Chr.1: 10430481-10430481 on Build GRCh38
rs941327180 Chr.1: 10430433-10430433 on Build GRCh38
rs914987060 Chr.1: 10430500-10430500 on Build GRCh38
rs982936032 Chr.1: 10430557-10430557 on Build GRCh38
rs966253737 Chr.1: 10430539-10430539 on Build GRCh38
rs1045392202 Chr.1: 10430445-10430445 on Build GRCh38
rs768705279 Chr.1: 10430581-10430581 on Build GRCh38
rs774882305 Chr.1: 10430594-10430594 on Build GRCh38
rs952711770 Chr.1: 10430423-10430423 on Build GRCh38
rs1007519312 Chr.1: 10430506-10430506 on Build GRCh38
rs1009591798 Chr.1: 10430486-10430486 on Build GRCh38
rs553739349 Chr.1: 10430461-10430461 on Build GRCh38
rs905086890 Chr.1: 10430455-10430455 on Build GRCh38
rs661272 Chr.1: 10430493-10430493 on Build GRCh38
rs746721788 Chr.1: 10430491-10430491 on Build GRCh38
rs1005981346 Chr.1: 10430430-10430430 on Build GRCh38

View Full Product Details