Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10395478-10395723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC37
Entrez Gene ID
11140
Gene Name
cell division cycle 37
Gene Aliases
P50CDC37
UniGene
Hs.160958
Gene Chromosome Location
Chr.19: 10391128-10403595 on Build GRCh38
Transcripts
NM_007065.3 XM_011527652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1158210 c.407C>T p.T136M Chr.19: 10395515-10395515 on Build GRCh38
5056170 c.388A>G p.N130D Chr.19: 10395534-10395534 on Build GRCh38
7234897 c.379-9C>G p.? Chr.19: 10395552-10395552 on Build GRCh38
6033073 c.428T>C p.V143A Chr.19: 10395494-10395494 on Build GRCh38
3735843 c.392C>T p.T131I Chr.19: 10395530-10395530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750419395 Chr.19: 10395606-10395606 on Build GRCh38
rs757882736 Chr.19: 10395610-10395610 on Build GRCh38
rs182055971 Chr.19: 10395523-10395523 on Build GRCh38
rs747081348 Chr.19: 10395644-10395644 on Build GRCh38
rs764233425 Chr.19: 10395679-10395680 on Build GRCh38
rs988144565 Chr.19: 10395512-10395512 on Build GRCh38
rs1041184270 Chr.19: 10395572-10395572 on Build GRCh38
rs757178328 Chr.19: 10395498-10395498 on Build GRCh38
rs761527270 Chr.19: 10395602-10395602 on Build GRCh38
rs774998656 Chr.19: 10395576-10395576 on Build GRCh38
rs931126458 Chr.19: 10395600-10395600 on Build GRCh38
rs768865378 Chr.19: 10395655-10395655 on Build GRCh38
rs577768027 Chr.19: 10395540-10395540 on Build GRCh38
rs751026992 Chr.19: 10395684-10395684 on Build GRCh38
rs746726788 Chr.19: 10395550-10395550 on Build GRCh38
rs942796988 Chr.19: 10395549-10395549 on Build GRCh38
rs1013993244 Chr.19: 10395687-10395687 on Build GRCh38
rs539822053 Chr.19: 10395641-10395641 on Build GRCh38
rs764253091 Chr.19: 10395599-10395599 on Build GRCh38
rs759681145 Chr.19: 10395584-10395584 on Build GRCh38
rs775590977 Chr.19: 10395619-10395620 on Build GRCh38
rs757758489 Chr.19: 10395626-10395626 on Build GRCh38
rs746423700 Chr.19: 10395640-10395640 on Build GRCh38
rs770026413 Chr.19: 10395556-10395556 on Build GRCh38
rs761583736 Chr.19: 10395666-10395666 on Build GRCh38
rs771511787 Chr.19: 10395565-10395565 on Build GRCh38
rs763095994 Chr.19: 10395675-10395676 on Build GRCh38
rs545904157 Chr.19: 10395546-10395546 on Build GRCh38
rs760995580 Chr.19: 10395596-10395596 on Build GRCh38
rs752346497 Chr.19: 10395704-10395704 on Build GRCh38
rs201068763 Chr.19: 10395560-10395560 on Build GRCh38
rs572827098 Chr.19: 10395632-10395632 on Build GRCh38
rs754454931 Chr.19: 10395685-10395685 on Build GRCh38
rs775597031 Chr.19: 10395643-10395643 on Build GRCh38
rs762840182 Chr.19: 10395681-10395681 on Build GRCh38
rs34379273 Chr.19: 10395559-10395560 on Build GRCh38
rs921184845 Chr.19: 10395561-10395561 on Build GRCh38
rs773104701 Chr.19: 10395678-10395678 on Build GRCh38
rs764523965 Chr.19: 10395585-10395585 on Build GRCh38
rs775550400 Chr.19: 10395591-10395591 on Build GRCh38
rs765155914 Chr.19: 10395605-10395605 on Build GRCh38
rs916934114 Chr.19: 10395705-10395705 on Build GRCh38
rs776785823 Chr.19: 10395664-10395664 on Build GRCh38
rs772333522 Chr.19: 10395587-10395587 on Build GRCh38
rs766253562 Chr.19: 10395683-10395683 on Build GRCh38
rs765066415 Chr.19: 10395668-10395668 on Build GRCh38
rs751065555 Chr.19: 10395614-10395614 on Build GRCh38
rs983934835 Chr.19: 10395617-10395617 on Build GRCh38
rs767190846 Chr.19: 10395694-10395694 on Build GRCh38
rs769795146 Chr.19: 10395609-10395610 on Build GRCh38
rs775106584 Chr.19: 10395649-10395649 on Build GRCh38
rs749604228 Chr.19: 10395618-10395618 on Build GRCh38
rs778842852 Chr.19: 10395522-10395522 on Build GRCh38
rs562012926 Chr.19: 10395682-10395682 on Build GRCh38
rs866708079 Chr.19: 10395690-10395690 on Build GRCh38

View Full Product Details