Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44058143-44058382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBNL
Entrez Gene ID
28988
Gene Name
drebrin like
Gene Aliases
ABP1, HIP-55, HIP55, SH3P7
UniGene
Hs.436500
Gene Chromosome Location
Chr.7: 44044640-44061716 on Build GRCh38
Transcripts
NM_001014436.2 NM_001122956.1 NM_001284313.1 NM_001284315.1 NM_014063.6 XM_011515325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6396889 c.634C>T p.R212C Chr.7: 44058210-44058210 on Build GRCh38
6418324 c.586C>T p.R196W Chr.7: 44058162-44058162 on Build GRCh38
4650753 c.626G>A p.R209H Chr.7: 44058202-44058202 on Build GRCh38
6379063 c.624G>T p.E208D Chr.7: 44058200-44058200 on Build GRCh38
6373941 c.589C>T p.R197W Chr.7: 44058165-44058165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759246763 Chr.7: 44058276-44058276 on Build GRCh38
rs770216157 Chr.7: 44058163-44058163 on Build GRCh38
rs757861014 Chr.7: 44058292-44058292 on Build GRCh38
rs770522281 Chr.7: 44058320-44058320 on Build GRCh38
rs754597320 Chr.7: 44058218-44058218 on Build GRCh38
rs752137757 Chr.7: 44058220-44058220 on Build GRCh38
rs374806538 Chr.7: 44058263-44058263 on Build GRCh38
rs748177361 Chr.7: 44058257-44058257 on Build GRCh38
rs770550443 Chr.7: 44058326-44058326 on Build GRCh38
rs1008498763 Chr.7: 44058182-44058182 on Build GRCh38
rs373414000 Chr.7: 44058204-44058204 on Build GRCh38
rs748349710 Chr.7: 44058326-44058326 on Build GRCh38
rs201466446 Chr.7: 44058246-44058246 on Build GRCh38
rs377760952 Chr.7: 44058206-44058206 on Build GRCh38
rs370249217 Chr.7: 44058219-44058219 on Build GRCh38
rs980627474 Chr.7: 44058226-44058226 on Build GRCh38
rs367760382 Chr.7: 44058232-44058232 on Build GRCh38
rs543782312 Chr.7: 44058234-44058234 on Build GRCh38
rs866677337 Chr.7: 44058235-44058235 on Build GRCh38
rs866293335 Chr.7: 44058211-44058211 on Build GRCh38
rs777527115 Chr.7: 44058184-44058184 on Build GRCh38
rs749568803 Chr.7: 44058239-44058239 on Build GRCh38
rs760232921 Chr.7: 44058312-44058312 on Build GRCh38
rs374954243 Chr.7: 44058231-44058231 on Build GRCh38
rs375372668 Chr.7: 44058362-44058362 on Build GRCh38
rs541074419 Chr.7: 44058264-44058264 on Build GRCh38
rs776391040 Chr.7: 44058275-44058275 on Build GRCh38
rs760569925 Chr.7: 44058265-44058265 on Build GRCh38
rs201356766 Chr.7: 44058165-44058165 on Build GRCh38
rs749482498 Chr.7: 44058170-44058170 on Build GRCh38
rs185229721 Chr.7: 44058357-44058357 on Build GRCh38
rs376675318 Chr.7: 44058244-44058244 on Build GRCh38
rs750934274 Chr.7: 44058302-44058302 on Build GRCh38
rs768897392 Chr.7: 44058171-44058171 on Build GRCh38
rs764867008 Chr.7: 44058278-44058278 on Build GRCh38
rs199569640 Chr.7: 44058205-44058205 on Build GRCh38
rs752328630 Chr.7: 44058291-44058291 on Build GRCh38
rs753525011 Chr.7: 44058210-44058210 on Build GRCh38
rs771873890 Chr.7: 44058162-44058162 on Build GRCh38
rs767354885 Chr.7: 44058202-44058202 on Build GRCh38
rs368233079 Chr.7: 44058313-44058313 on Build GRCh38
rs766206080 Chr.7: 44058272-44058272 on Build GRCh38
rs756665569 Chr.7: 44058310-44058310 on Build GRCh38
rs772640287 Chr.7: 44058309-44058310 on Build GRCh38
rs757851309 Chr.7: 44058230-44058230 on Build GRCh38
rs529781892 Chr.7: 44058348-44058348 on Build GRCh38
rs774552738 Chr.7: 44058175-44058175 on Build GRCh38
rs547969086 Chr.7: 44058354-44058354 on Build GRCh38
rs961747230 Chr.7: 44058192-44058192 on Build GRCh38
rs946286440 Chr.7: 44058356-44058356 on Build GRCh38
rs559022415 Chr.7: 44058324-44058324 on Build GRCh38
rs761870389 Chr.7: 44058183-44058183 on Build GRCh38
rs763610905 Chr.7: 44058299-44058299 on Build GRCh38

View Full Product Details