Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 11629364-11629619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
UniGene
Hs.567259
Gene Chromosome Location
Chr.17: 11598431-11970168 on Build GRCh38
Transcripts
NM_001372.3 XM_011523703.1 XM_017024292.1 XM_017024294.1 XM_017024295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1168828 c.1408G>A p.V470I Chr.17: 11629474-11629474 on Build GRCh38
6434575 c.1508T>C p.L503P Chr.17: 11629574-11629574 on Build GRCh38
3514050 c.1450G>A p.E484K Chr.17: 11629516-11629516 on Build GRCh38
6900866 c.1369C>T p.L457L Chr.17: 11629435-11629435 on Build GRCh38
6646978 c.1441C>T p.Q481* Chr.17: 11629507-11629507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760092268 Chr.17: 11629548-11629548 on Build GRCh38
rs377142502 Chr.17: 11629444-11629444 on Build GRCh38
rs756414051 Chr.17: 11629497-11629497 on Build GRCh38
rs1009109182 Chr.17: 11629565-11629565 on Build GRCh38
rs978699929 Chr.17: 11629436-11629436 on Build GRCh38
rs770867759 Chr.17: 11629501-11629501 on Build GRCh38
rs758976488 Chr.17: 11629489-11629490 on Build GRCh38
rs1050087529 Chr.17: 11629543-11629543 on Build GRCh38
rs369047155 Chr.17: 11629523-11629523 on Build GRCh38
rs149806029 Chr.17: 11629427-11629427 on Build GRCh38
rs372072983 Chr.17: 11629512-11629512 on Build GRCh38
rs78960618 Chr.17: 11629555-11629555 on Build GRCh38
rs770553421 Chr.17: 11629409-11629409 on Build GRCh38
rs181219739 Chr.17: 11629403-11629403 on Build GRCh38
rs751634130 Chr.17: 11629473-11629473 on Build GRCh38
rs369440707 Chr.17: 11629452-11629452 on Build GRCh38
rs774961046 Chr.17: 11629430-11629430 on Build GRCh38
rs73290804 Chr.17: 11629470-11629470 on Build GRCh38
rs778178016 Chr.17: 11629580-11629580 on Build GRCh38
rs776724665 Chr.17: 11629534-11629534 on Build GRCh38
rs200171758 Chr.17: 11629474-11629474 on Build GRCh38
rs752879194 Chr.17: 11629561-11629561 on Build GRCh38
rs374206604 Chr.17: 11629560-11629560 on Build GRCh38
rs570408216 Chr.17: 11629441-11629441 on Build GRCh38
rs759011256 Chr.17: 11629421-11629421 on Build GRCh38
rs751970112 Chr.17: 11629547-11629547 on Build GRCh38
rs748314461 Chr.17: 11629494-11629494 on Build GRCh38
rs766788032 Chr.17: 11629541-11629541 on Build GRCh38
rs559139037 Chr.17: 11629471-11629471 on Build GRCh38
rs749159702 Chr.17: 11629498-11629498 on Build GRCh38
rs148518864 Chr.17: 11629479-11629479 on Build GRCh38
rs774990587 Chr.17: 11629517-11629517 on Build GRCh38
rs142794912 Chr.17: 11629508-11629508 on Build GRCh38
rs772061043 Chr.17: 11629595-11629595 on Build GRCh38
rs113665087 Chr.17: 11629463-11629463 on Build GRCh38
rs745843806 Chr.17: 11629511-11629511 on Build GRCh38
rs771447371 Chr.17: 11629514-11629514 on Build GRCh38
rs756397601 Chr.17: 11629570-11629570 on Build GRCh38
rs773905856 Chr.17: 11629409-11629409 on Build GRCh38
rs141702885 Chr.17: 11629583-11629583 on Build GRCh38
rs200240255 Chr.17: 11629540-11629540 on Build GRCh38
rs868374603 Chr.17: 11629550-11629550 on Build GRCh38
rs778802993 Chr.17: 11629588-11629588 on Build GRCh38
rs9895535 Chr.17: 11629518-11629518 on Build GRCh38
rs773979808 Chr.17: 11629461-11629463 on Build GRCh38
rs28665591 Chr.17: 11629398-11629398 on Build GRCh38

View Full Product Details