Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90726414-90726675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK14
Entrez Gene ID
5218
Gene Name
cyclin dependent kinase 14
Gene Aliases
PFTAIRE1, PFTK1
UniGene
Hs.258576
Gene Chromosome Location
Chr.7: 90595994-91210590 on Build GRCh38
Transcripts
NM_001287135.1 NM_001287136.1 NM_001287137.1 NM_012395.3 XM_005250436.3 XM_005250438.3 XM_005250439.2 XM_011516306.2 XM_011516307.2 XM_017012320.1 XM_017012321.1 XM_017012322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6536293 c.155A>G p.K52R Chr.7: 90726652-90726652 on Build GRCh38
1092832 c.134A>G p.D45G Chr.7: 90726631-90726631 on Build GRCh38
6447518 c.131T>C p.V44A Chr.7: 90726574-90726574 on Build GRCh38
1488764 c.145G>A p.E49K Chr.7: 90726642-90726642 on Build GRCh38
6625384 c.88T>C p.S30P Chr.7: 90726585-90726585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759722377 Chr.7: 90726606-90726606 on Build GRCh38
rs770216476 Chr.7: 90726575-90726575 on Build GRCh38
rs550106751 Chr.7: 90726554-90726554 on Build GRCh38
rs144891494 Chr.7: 90726626-90726626 on Build GRCh38
rs1034377957 Chr.7: 90726505-90726505 on Build GRCh38
rs781484660 Chr.7: 90726552-90726554 on Build GRCh38
rs763175488 Chr.7: 90726591-90726591 on Build GRCh38
rs745973909 Chr.7: 90726584-90726584 on Build GRCh38
rs983642763 Chr.7: 90726627-90726627 on Build GRCh38
rs748667177 Chr.7: 90726572-90726572 on Build GRCh38
rs773595715 Chr.7: 90726590-90726590 on Build GRCh38
rs371519163 Chr.7: 90726528-90726528 on Build GRCh38
rs774434672 Chr.7: 90726601-90726601 on Build GRCh38
rs959594994 Chr.7: 90726567-90726567 on Build GRCh38
rs140679817 Chr.7: 90726592-90726592 on Build GRCh38
rs767512259 Chr.7: 90726617-90726617 on Build GRCh38
rs765218989 Chr.7: 90726639-90726639 on Build GRCh38
rs150323625 Chr.7: 90726501-90726501 on Build GRCh38
rs746567651 Chr.7: 90726520-90726520 on Build GRCh38
rs775644986 Chr.7: 90726624-90726624 on Build GRCh38
rs763177842 Chr.7: 90726480-90726480 on Build GRCh38
rs182257443 Chr.7: 90726548-90726548 on Build GRCh38
rs973588185 Chr.7: 90726581-90726581 on Build GRCh38

View Full Product Details