Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19024044-19024295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAP
Entrez Gene ID
10750
Gene Name
GRB2-related adaptor protein
Gene Aliases
-
UniGene
Hs.567416
Gene Chromosome Location
Chr.17: 19020656-19051352 on Build GRCh38
Transcripts
NM_006613.3 XM_017024029.1 XM_017024030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7302639 c.447C>T p.R149R Chr.17: 19024236-19024236 on Build GRCh38
3969871 c.420C>T p.I140I Chr.17: 19024263-19024263 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs867944921 Chr.17: 19024120-19024120 on Build GRCh38
rs376195291 Chr.17: 19024067-19024067 on Build GRCh38
rs200201794 Chr.17: 19024254-19024254 on Build GRCh38
rs764488447 Chr.17: 19024275-19024275 on Build GRCh38
rs764725994 Chr.17: 19024197-19024197 on Build GRCh38
rs143302839 Chr.17: 19024256-19024256 on Build GRCh38
rs546369217 Chr.17: 19024145-19024145 on Build GRCh38
rs371370322 Chr.17: 19024236-19024236 on Build GRCh38
rs763340856 Chr.17: 19024184-19024184 on Build GRCh38
rs779652857 Chr.17: 19024204-19024204 on Build GRCh38
rs775710004 Chr.17: 19024270-19024270 on Build GRCh38
rs562078975 Chr.17: 19024271-19024271 on Build GRCh38
rs763370391 Chr.17: 19024194-19024194 on Build GRCh38
rs771901263 Chr.17: 19024237-19024237 on Build GRCh38
rs752148832 Chr.17: 19024276-19024276 on Build GRCh38
rs377726893 Chr.17: 19024269-19024269 on Build GRCh38
rs948380177 Chr.17: 19024178-19024178 on Build GRCh38
rs143281137 Chr.17: 19024235-19024235 on Build GRCh38
rs749889999 Chr.17: 19024200-19024200 on Build GRCh38
rs560122120 Chr.17: 19024164-19024164 on Build GRCh38
rs755672965 Chr.17: 19024203-19024203 on Build GRCh38
rs369543027 Chr.17: 19024066-19024066 on Build GRCh38
rs759847345 Chr.17: 19024257-19024257 on Build GRCh38
rs928382002 Chr.17: 19024206-19024206 on Build GRCh38
rs962951190 Chr.17: 19024108-19024108 on Build GRCh38
rs778587592 Chr.17: 19024232-19024232 on Build GRCh38
rs753186814 Chr.17: 19024190-19024190 on Build GRCh38
rs868525777 Chr.17: 19024214-19024214 on Build GRCh38
rs543982462 Chr.17: 19024098-19024098 on Build GRCh38
rs554964941 Chr.17: 19024082-19024082 on Build GRCh38
rs747053384 Chr.17: 19024250-19024250 on Build GRCh38
rs765715084 Chr.17: 19024165-19024165 on Build GRCh38
rs557786542 Chr.17: 19024126-19024126 on Build GRCh38
rs777812775 Chr.17: 19024238-19024238 on Build GRCh38
rs770213883 Chr.17: 19024262-19024262 on Build GRCh38
rs577410357 Chr.17: 19024135-19024135 on Build GRCh38
rs535108648 Chr.17: 19024072-19024072 on Build GRCh38
rs771164685 Chr.17: 19024252-19024252 on Build GRCh38
rs565247493 Chr.17: 19024083-19024083 on Build GRCh38
rs199803859 Chr.17: 19024231-19024231 on Build GRCh38
rs1015883209 Chr.17: 19024099-19024099 on Build GRCh38
rs754719809 Chr.17: 19024221-19024221 on Build GRCh38
rs753525042 Chr.17: 19024217-19024217 on Build GRCh38
rs9893211 Chr.17: 19024233-19024233 on Build GRCh38
rs1029715782 Chr.17: 19024076-19024076 on Build GRCh38

View Full Product Details