Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141915867-141916132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B3
Entrez Gene ID
483
Gene Name
ATPase Na+/K+ transporting subunit beta 3
Gene Aliases
ATPB-3, CD298
UniGene
Hs.477789
Gene Chromosome Location
Chr.3: 141876628-141926540 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3124858 c.567A>G p.I189M Chr.3: 141916005-141916005 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987047509 Chr.3: 141916061-141916061 on Build GRCh38
rs553141902 Chr.3: 141916027-141916027 on Build GRCh38
rs369684231 Chr.3: 141915933-141915933 on Build GRCh38
rs952507213 Chr.3: 141915984-141915984 on Build GRCh38
rs746519364 Chr.3: 141916053-141916053 on Build GRCh38
rs776810302 Chr.3: 141916083-141916083 on Build GRCh38
rs929839673 Chr.3: 141916099-141916099 on Build GRCh38
rs573414606 Chr.3: 141916095-141916095 on Build GRCh38
rs768110412 Chr.3: 141916055-141916055 on Build GRCh38
rs764986305 Chr.3: 141915930-141915930 on Build GRCh38
rs776304575 Chr.3: 141916057-141916057 on Build GRCh38
rs762744492 Chr.3: 141916071-141916071 on Build GRCh38
rs961503682 Chr.3: 141916075-141916075 on Build GRCh38
rs776214491 Chr.3: 141915928-141915928 on Build GRCh38
rs868437047 Chr.3: 141916033-141916033 on Build GRCh38
rs761626729 Chr.3: 141915929-141915929 on Build GRCh38
rs535010007 Chr.3: 141915936-141915936 on Build GRCh38
rs749868355 Chr.3: 141916038-141916038 on Build GRCh38
rs755027319 Chr.3: 141915949-141915949 on Build GRCh38
rs747775692 Chr.3: 141916058-141916058 on Build GRCh38
rs913516162 Chr.3: 141915988-141915988 on Build GRCh38
rs757705858 Chr.3: 141916041-141916041 on Build GRCh38
rs373593164 Chr.3: 141916044-141916044 on Build GRCh38
rs367701851 Chr.3: 141916068-141916068 on Build GRCh38
rs768306587 Chr.3: 141915925-141915925 on Build GRCh38
rs751597693 Chr.3: 141915941-141915941 on Build GRCh38
rs778263681 Chr.3: 141916036-141916036 on Build GRCh38
rs988971996 Chr.3: 141915939-141915939 on Build GRCh38
rs147932119 Chr.3: 141915994-141915994 on Build GRCh38
rs756591629 Chr.3: 141916034-141916034 on Build GRCh38
rs767875630 Chr.3: 141915996-141915996 on Build GRCh38
rs1012564071 Chr.3: 141915922-141915922 on Build GRCh38
rs762785935 Chr.3: 141915934-141915934 on Build GRCh38
rs969744138 Chr.3: 141915894-141915894 on Build GRCh38
rs192303297 Chr.3: 141916098-141916098 on Build GRCh38
rs578129421 Chr.3: 141916063-141916063 on Build GRCh38

View Full Product Details