Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247565918-247566153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCSAML
Entrez Gene ID
148823
Gene Name
germinal center associated signaling and motility like
Gene Aliases
C1orf150
UniGene
Hs.732113
Gene Chromosome Location
Chr.1: 247507058-247577690 on Build GRCh38
Transcripts
NM_001281834.1 NM_001281835.1 NM_001281836.1 NM_001281837.1 NM_001281838.1 NM_001281853.1 NM_145278.4 XM_017000399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
388448 c.161C>G p.S54C Chr.1: 247565952-247565952 on Build GRCh38
5380369 c.141C>T p.S47S Chr.1: 247565932-247565932 on Build GRCh38
907043 c.147A>C p.E49D Chr.1: 247565938-247565938 on Build GRCh38
3785223 c.146A>T p.E49V Chr.1: 247565937-247565937 on Build GRCh38
231890 c.152C>T p.S51L Chr.1: 247565943-247565943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770382758 Chr.1: 247565945-247565945 on Build GRCh38
rs955809269 Chr.1: 247566040-247566040 on Build GRCh38
rs12141535 Chr.1: 247566089-247566089 on Build GRCh38
rs373404059 Chr.1: 247565948-247565948 on Build GRCh38
rs914700297 Chr.1: 247566093-247566093 on Build GRCh38
rs924850506 Chr.1: 247566117-247566117 on Build GRCh38
rs775117323 Chr.1: 247565965-247565965 on Build GRCh38
rs762340218 Chr.1: 247565980-247565980 on Build GRCh38
rs978003518 Chr.1: 247566110-247566110 on Build GRCh38
rs971053171 Chr.1: 247566051-247566051 on Build GRCh38
rs745356771 Chr.1: 247565949-247565949 on Build GRCh38
rs12141536 Chr.1: 247566098-247566098 on Build GRCh38
rs1002803144 Chr.1: 247566062-247566062 on Build GRCh38
rs376815601 Chr.1: 247565978-247565978 on Build GRCh38
rs115161475 Chr.1: 247566125-247566125 on Build GRCh38
rs765640694 Chr.1: 247566000-247566000 on Build GRCh38
rs188761411 Chr.1: 247566024-247566024 on Build GRCh38
rs548732030 Chr.1: 247566009-247566009 on Build GRCh38
rs375598595 Chr.1: 247566001-247566001 on Build GRCh38
rs369694422 Chr.1: 247566021-247566021 on Build GRCh38
rs531352871 Chr.1: 247566026-247566026 on Build GRCh38
rs1035663899 Chr.1: 247566078-247566078 on Build GRCh38
rs1017490467 Chr.1: 247565990-247565990 on Build GRCh38
rs771747638 Chr.1: 247565957-247565957 on Build GRCh38
rs549850676 Chr.1: 247566087-247566087 on Build GRCh38
rs1015475872 Chr.1: 247566028-247566028 on Build GRCh38
rs761084882 Chr.1: 247565973-247565973 on Build GRCh38
rs766768093 Chr.1: 247566010-247566010 on Build GRCh38

View Full Product Details