Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87317035-87317294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B13
Entrez Gene ID
345275
Gene Name
hydroxysteroid 17-beta dehydrogenase 13
Gene Aliases
HMFN0376, NIIL497, SCDR9, SDR16C3
UniGene
Hs.284414
Gene Chromosome Location
Chr.4: 87303789-87322906 on Build GRCh38
Transcripts
NM_001136230.2 NM_178135.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5017484 c.448delT p.W150fs*10 Chr.4: 87317094-87317094 on Build GRCh38
4138540 c.450G>T p.W150C Chr.4: 87317092-87317092 on Build GRCh38
5355943 c.320T>C p.V107A Chr.4: 87317222-87317222 on Build GRCh38
3606825 c.428T>C p.V143A Chr.4: 87317114-87317114 on Build GRCh38
1431587 c.382G>A p.D128N Chr.4: 87317160-87317160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547893464 Chr.4: 87317256-87317257 on Build GRCh38
rs767391751 Chr.4: 87317095-87317095 on Build GRCh38
rs564292392 Chr.4: 87317233-87317233 on Build GRCh38
rs757620391 Chr.4: 87317188-87317188 on Build GRCh38
rs763275950 Chr.4: 87317077-87317077 on Build GRCh38
rs779627733 Chr.4: 87317160-87317160 on Build GRCh38
rs745984072 Chr.4: 87317063-87317063 on Build GRCh38
rs61748262 Chr.4: 87317092-87317092 on Build GRCh38
rs528603881 Chr.4: 87317264-87317264 on Build GRCh38
rs372936841 Chr.4: 87317161-87317161 on Build GRCh38
rs753277334 Chr.4: 87317147-87317147 on Build GRCh38
rs866207827 Chr.4: 87317242-87317242 on Build GRCh38
rs142525792 Chr.4: 87317104-87317104 on Build GRCh38
rs374101728 Chr.4: 87317068-87317068 on Build GRCh38
rs746420270 Chr.4: 87317193-87317193 on Build GRCh38
rs761775014 Chr.4: 87317181-87317182 on Build GRCh38
rs768944122 Chr.4: 87317083-87317083 on Build GRCh38
rs765903026 Chr.4: 87317145-87317145 on Build GRCh38
rs190310719 Chr.4: 87317194-87317194 on Build GRCh38
rs749370188 Chr.4: 87317212-87317212 on Build GRCh38
rs751935394 Chr.4: 87317173-87317173 on Build GRCh38
rs371923490 Chr.4: 87317239-87317239 on Build GRCh38
rs904902676 Chr.4: 87317094-87317094 on Build GRCh38
rs372816850 Chr.4: 87317211-87317211 on Build GRCh38
rs771920377 Chr.4: 87317247-87317247 on Build GRCh38
rs201763274 Chr.4: 87317141-87317141 on Build GRCh38
rs370289965 Chr.4: 87317085-87317085 on Build GRCh38
rs778776813 Chr.4: 87317196-87317196 on Build GRCh38
rs760664521 Chr.4: 87317253-87317253 on Build GRCh38
rs376964857 Chr.4: 87317254-87317254 on Build GRCh38

View Full Product Details