Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 60460214-60460483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL11A
Entrez Gene ID
53335
Gene Name
B-cell CLL/lymphoma 11A
Gene Aliases
BCL11A-L, BCL11A-S, BCL11A-XL, BCL11a-M, CTIP1, EVI9, HBFQTL5, ZNF856
UniGene
Hs.370549
Gene Chromosome Location
Chr.2: 60451167-60553498 on Build GRCh38
Transcripts
NM_018014.3 NM_022893.3 NM_138559.1 XM_011532909.1 XM_011532910.1 XM_011532912.1 XM_017004333.1 XM_017004334.1 XM_017004335.1 XM_017004336.1 XM_017004337.1 XM_017004338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
218729 c.2431G>A p.V811M Chr.2: 60460481-60460481 on Build GRCh38
3659911 c.2484G>T p.L828F Chr.2: 60460428-60460428 on Build GRCh38
722214 c.2429G>T p.S810I Chr.2: 60460483-60460483 on Build GRCh38
6994540 c.2473G>T p.D825Y Chr.2: 60460439-60460439 on Build GRCh38
5726892 c.2481G>A p.V827V Chr.2: 60460431-60460431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779663540 Chr.2: 60460389-60460389 on Build GRCh38
rs943281672 Chr.2: 60460383-60460383 on Build GRCh38
rs534998425 Chr.2: 60460271-60460271 on Build GRCh38
rs1008925659 Chr.2: 60460464-60460464 on Build GRCh38
rs753840883 Chr.2: 60460400-60460400 on Build GRCh38
rs970307742 Chr.2: 60460258-60460258 on Build GRCh38
rs767054274 Chr.2: 60460387-60460387 on Build GRCh38
rs945187909 Chr.2: 60460365-60460365 on Build GRCh38
rs767573848 Chr.2: 60460266-60460269 on Build GRCh38
rs989290040 Chr.2: 60460304-60460304 on Build GRCh38
rs890452152 Chr.2: 60460416-60460416 on Build GRCh38
rs749992547 Chr.2: 60460388-60460388 on Build GRCh38
rs754806293 Chr.2: 60460418-60460418 on Build GRCh38
rs771597942 Chr.2: 60460436-60460436 on Build GRCh38
rs957428250 Chr.2: 60460286-60460286 on Build GRCh38
rs778837788 Chr.2: 60460435-60460435 on Build GRCh38
rs760723370 Chr.2: 60460361-60460361 on Build GRCh38
rs779121016 Chr.2: 60460357-60460358 on Build GRCh38
rs898817151 Chr.2: 60460401-60460401 on Build GRCh38
rs913341742 Chr.2: 60460321-60460321 on Build GRCh38
rs761584158 Chr.2: 60460377-60460377 on Build GRCh38
rs777875486 Chr.2: 60460269-60460270 on Build GRCh38
rs747406096 Chr.2: 60460322-60460322 on Build GRCh38
rs759898984 Chr.2: 60460371-60460371 on Build GRCh38
rs770956284 Chr.2: 60460362-60460362 on Build GRCh38
rs35310820 Chr.2: 60460366-60460366 on Build GRCh38
rs538290922 Chr.2: 60460244-60460244 on Build GRCh38
rs946516901 Chr.2: 60460243-60460243 on Build GRCh38
rs761630332 Chr.2: 60460349-60460349 on Build GRCh38
rs765634143 Chr.2: 60460373-60460373 on Build GRCh38
rs752998646 Chr.2: 60460376-60460376 on Build GRCh38
rs932013583 Chr.2: 60460364-60460364 on Build GRCh38
rs553161595 Chr.2: 60460356-60460357 on Build GRCh38
rs371406296 Chr.2: 60460369-60460369 on Build GRCh38
rs373524493 Chr.2: 60460390-60460390 on Build GRCh38
rs201706698 Chr.2: 60460265-60460266 on Build GRCh38

View Full Product Details