Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52465350-52465603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2
Entrez Gene ID
26586
Gene Name
cytoskeleton associated protein 2
Gene Aliases
LB1, TMAP, se20-10
UniGene
Hs.444028
Gene Chromosome Location
Chr.13: 52455360-52476631 on Build GRCh38
Transcripts
NM_001098525.2 NM_001286686.1 NM_001286687.1 NM_018204.4 XM_005266344.4 XM_011535043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6139038 c.1456G>A p.A486T Chr.13: 52465445-52465445 on Build GRCh38
4903181 c.1424C>T p.P475L Chr.13: 52465413-52465413 on Build GRCh38
5118016 c.1476+7T>C p.? Chr.13: 52465472-52465472 on Build GRCh38
1943443 c.1449T>C p.Y483Y Chr.13: 52465438-52465438 on Build GRCh38
1943444 c.1473T>A p.A491A Chr.13: 52465462-52465462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371952071 Chr.13: 52465515-52465515 on Build GRCh38
rs374856873 Chr.13: 52465495-52465495 on Build GRCh38
rs773311360 Chr.13: 52465470-52465470 on Build GRCh38
rs143514154 Chr.13: 52465412-52465412 on Build GRCh38
rs763680534 Chr.13: 52465415-52465415 on Build GRCh38
rs114093183 Chr.13: 52465493-52465493 on Build GRCh38
rs775355265 Chr.13: 52465513-52465513 on Build GRCh38
rs934878662 Chr.13: 52465544-52465544 on Build GRCh38
rs760931039 Chr.13: 52465383-52465383 on Build GRCh38
rs755402568 Chr.13: 52465463-52465463 on Build GRCh38
rs766552497 Chr.13: 52465390-52465390 on Build GRCh38
rs779526094 Chr.13: 52465464-52465464 on Build GRCh38
rs776732770 Chr.13: 52465494-52465494 on Build GRCh38
rs367879017 Chr.13: 52465498-52465498 on Build GRCh38
rs374510275 Chr.13: 52465468-52465468 on Build GRCh38
rs763976569 Chr.13: 52465516-52465516 on Build GRCh38
rs201479437 Chr.13: 52465395-52465395 on Build GRCh38
rs765047738 Chr.13: 52465512-52465512 on Build GRCh38
rs748690487 Chr.13: 52465466-52465466 on Build GRCh38
rs757796870 Chr.13: 52465423-52465425 on Build GRCh38
rs757862089 Chr.13: 52465413-52465413 on Build GRCh38
rs545573962 Chr.13: 52465548-52465548 on Build GRCh38
rs780770276 Chr.13: 52465443-52465443 on Build GRCh38
rs61959701 Chr.13: 52465506-52465506 on Build GRCh38
rs765100845 Chr.13: 52465406-52465406 on Build GRCh38
rs895953765 Chr.13: 52465566-52465566 on Build GRCh38
rs201547657 Chr.13: 52465472-52465472 on Build GRCh38
rs759384590 Chr.13: 52465400-52465400 on Build GRCh38
rs749653423 Chr.13: 52465454-52465454 on Build GRCh38
rs756900004 Chr.13: 52465436-52465436 on Build GRCh38
rs757506861 Chr.13: 52465379-52465379 on Build GRCh38
rs751076159 Chr.13: 52465425-52465425 on Build GRCh38
rs903451356 Chr.13: 52465527-52465527 on Build GRCh38

View Full Product Details