Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45518733-45519006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
UniGene
Hs.24601
Gene Chromosome Location
Chr.22: 45502839-45601134 on Build GRCh38
Transcripts
NM_001996.3 NM_006485.3 NM_006486.2 NM_006487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74622 c.149G>A p.C50Y Chr.22: 45518751-45518751 on Build GRCh38
1035071 c.178G>T p.E60* Chr.22: 45518780-45518780 on Build GRCh38
1035068 c.152C>G p.S51W Chr.22: 45518754-45518754 on Build GRCh38
3912971 c.152C>T p.S51L Chr.22: 45518754-45518754 on Build GRCh38
7325083 c.136C>A p.H46N Chr.22: 45518738-45518738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942959653 Chr.22: 45518796-45518796 on Build GRCh38
rs775783524 Chr.22: 45518804-45518804 on Build GRCh38
rs771919897 Chr.22: 45518806-45518818 on Build GRCh38
rs776003755 Chr.22: 45518755-45518755 on Build GRCh38
rs537556633 Chr.22: 45518761-45518761 on Build GRCh38
rs772445995 Chr.22: 45518754-45518754 on Build GRCh38
rs773071799 Chr.22: 45518827-45518828 on Build GRCh38
rs778486857 Chr.22: 45518799-45518799 on Build GRCh38
rs1010966363 Chr.22: 45518900-45518900 on Build GRCh38
rs899018937 Chr.22: 45518858-45518858 on Build GRCh38
rs764230125 Chr.22: 45518769-45518769 on Build GRCh38
rs1008685287 Chr.22: 45518949-45518959 on Build GRCh38
rs571562360 Chr.22: 45518936-45518936 on Build GRCh38
rs942840802 Chr.22: 45518972-45518972 on Build GRCh38
rs369617011 Chr.22: 45518833-45518833 on Build GRCh38
rs976879572 Chr.22: 45518980-45518980 on Build GRCh38
rs565538668 Chr.22: 45518850-45518850 on Build GRCh38
rs911412276 Chr.22: 45518969-45518969 on Build GRCh38
rs764553254 Chr.22: 45518902-45518902 on Build GRCh38
rs762072523 Chr.22: 45518775-45518775 on Build GRCh38
rs993975670 Chr.22: 45518867-45518867 on Build GRCh38
rs78488153 Chr.22: 45518795-45518795 on Build GRCh38
rs913674090 Chr.22: 45518983-45518986 on Build GRCh38
rs112698995 Chr.22: 45518917-45518917 on Build GRCh38
rs77035925 Chr.22: 45518812-45518812 on Build GRCh38
rs961685211 Chr.22: 45518756-45518756 on Build GRCh38
rs763181719 Chr.22: 45518857-45518857 on Build GRCh38
rs147045275 Chr.22: 45518791-45518791 on Build GRCh38
rs537370861 Chr.22: 45518957-45518957 on Build GRCh38
rs985883931 Chr.22: 45518968-45518968 on Build GRCh38
rs767425502 Chr.22: 45518788-45518788 on Build GRCh38
rs772352539 Chr.22: 45518803-45518803 on Build GRCh38
rs1003454325 Chr.22: 45518927-45518927 on Build GRCh38
rs557099204 Chr.22: 45518976-45518976 on Build GRCh38
rs375402034 Chr.22: 45518792-45518792 on Build GRCh38
rs905027849 Chr.22: 45518923-45518923 on Build GRCh38
rs557342504 Chr.22: 45518918-45518918 on Build GRCh38
rs745529980 Chr.22: 45518800-45518800 on Build GRCh38
rs971719152 Chr.22: 45518949-45518949 on Build GRCh38
rs1052412979 Chr.22: 45518886-45518886 on Build GRCh38
rs867191908 Chr.22: 45518771-45518771 on Build GRCh38
rs76022310 Chr.22: 45518876-45518876 on Build GRCh38
rs765517810 Chr.22: 45518778-45518778 on Build GRCh38
rs750698050 Chr.22: 45518786-45518786 on Build GRCh38
rs762438069 Chr.22: 45518965-45518965 on Build GRCh38
rs751982304 Chr.22: 45518964-45518964 on Build GRCh38
rs763043444 Chr.22: 45518797-45518797 on Build GRCh38
rs754762089 Chr.22: 45518787-45518787 on Build GRCh38
rs756788965 Chr.22: 45518797-45518797 on Build GRCh38
rs747538164 Chr.22: 45518808-45518808 on Build GRCh38
rs147635694 Chr.22: 45518834-45518834 on Build GRCh38
rs893894595 Chr.22: 45518887-45518887 on Build GRCh38
rs752679919 Chr.22: 45518790-45518790 on Build GRCh38
rs776630676 Chr.22: 45518770-45518770 on Build GRCh38
rs867934033 Chr.22: 45518843-45518843 on Build GRCh38
rs922856602 Chr.22: 45518984-45518984 on Build GRCh38
rs761090177 Chr.22: 45518764-45518764 on Build GRCh38
rs1034905079 Chr.22: 45518931-45518931 on Build GRCh38

View Full Product Details