Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24318721-24318958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF9
Entrez Gene ID
338872
Gene Name
C1q and tumor necrosis factor related protein 9
Gene Aliases
AQL1, C1QTNF9A, CTRP9
UniGene
Hs.362854
Gene Chromosome Location
Chr.13: 24307166-24322531 on Build GRCh38
Transcripts
NM_001303137.1 NM_001303138.1 NM_178540.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6479465 c.167-1G>T p.? Chr.13: 24318817-24318817 on Build GRCh38
3467802 c.221G>A p.G74E Chr.13: 24318872-24318872 on Build GRCh38
6875932 c.196A>G p.K66E Chr.13: 24318847-24318847 on Build GRCh38
469260 c.221G>T p.G74V Chr.13: 24318872-24318872 on Build GRCh38
1513919 c.227G>A p.R76Q Chr.13: 24318878-24318878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759260616 Chr.13: 24318881-24318882 on Build GRCh38
rs748534982 Chr.13: 24318881-24318881 on Build GRCh38
rs749817437 Chr.13: 24318794-24318794 on Build GRCh38
rs760985100 Chr.13: 24318885-24318885 on Build GRCh38
rs941164539 Chr.13: 24318811-24318811 on Build GRCh38
rs764196906 Chr.13: 24318916-24318916 on Build GRCh38
rs573399680 Chr.13: 24318812-24318812 on Build GRCh38
rs546281490 Chr.13: 24318801-24318801 on Build GRCh38
rs780226763 Chr.13: 24318793-24318793 on Build GRCh38
rs150781719 Chr.13: 24318829-24318829 on Build GRCh38
rs775479111 Chr.13: 24318901-24318901 on Build GRCh38
rs369453137 Chr.13: 24318888-24318888 on Build GRCh38
rs751174829 Chr.13: 24318833-24318833 on Build GRCh38
rs181829504 Chr.13: 24318804-24318804 on Build GRCh38
rs749630211 Chr.13: 24318858-24318858 on Build GRCh38
rs772248572 Chr.13: 24318803-24318803 on Build GRCh38
rs752497059 Chr.13: 24318823-24318823 on Build GRCh38
rs997580486 Chr.13: 24318900-24318900 on Build GRCh38
rs769112380 Chr.13: 24318799-24318799 on Build GRCh38
rs549791788 Chr.13: 24318921-24318921 on Build GRCh38
rs761867840 Chr.13: 24318802-24318802 on Build GRCh38
rs746288436 Chr.13: 24318787-24318787 on Build GRCh38
rs144539792 Chr.13: 24318842-24318842 on Build GRCh38
rs773114158 Chr.13: 24318884-24318884 on Build GRCh38
rs375254759 Chr.13: 24318791-24318791 on Build GRCh38
rs765098049 Chr.13: 24318813-24318813 on Build GRCh38
rs762832591 Chr.13: 24318906-24318906 on Build GRCh38
rs761599171 Chr.13: 24318922-24318922 on Build GRCh38
rs142688497 Chr.13: 24318877-24318877 on Build GRCh38
rs188339290 Chr.13: 24318780-24318780 on Build GRCh38
rs149288885 Chr.13: 24318830-24318830 on Build GRCh38
rs756913251 Chr.13: 24318767-24318767 on Build GRCh38
rs756789682 Chr.13: 24318837-24318837 on Build GRCh38
rs780428044 Chr.13: 24318857-24318857 on Build GRCh38
rs78731266 Chr.13: 24318843-24318843 on Build GRCh38
rs755673923 Chr.13: 24318849-24318849 on Build GRCh38
rs779386798 Chr.13: 24318878-24318878 on Build GRCh38

View Full Product Details