Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 96384234-96384459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC82
Entrez Gene ID
79780
Gene Name
coiled-coil domain containing 82
Gene Aliases
HSPC048
UniGene
Hs.525088
Gene Chromosome Location
Chr.11: 96352765-96389940 on Build GRCh38
Transcripts
NM_001318736.1 NM_001318737.1 NM_024725.3 XM_005274292.3 XM_006718911.3 XM_011542984.2 XM_011542987.2 XM_011542988.2 XM_017018305.1 XM_017018306.1 XM_017018307.1 XM_017018308.1 XM_017018309.1 XM_017018310.1 XM_017018311.1 XM_017018312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5637831 c.368T>A p.L123* Chr.11: 96384380-96384380 on Build GRCh38
467616 c.437A>G p.E146G Chr.11: 96384311-96384311 on Build GRCh38
6622501 c.384delA p.K128fs*3 Chr.11: 96384364-96384364 on Build GRCh38
1605417 c.316G>T p.G106C Chr.11: 96384432-96384432 on Build GRCh38
430122 c.390A>T p.L130F Chr.11: 96384358-96384358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773304204 Chr.11: 96384410-96384410 on Build GRCh38
rs766660190 Chr.11: 96384315-96384315 on Build GRCh38
rs771113136 Chr.11: 96384432-96384432 on Build GRCh38
rs117071588 Chr.11: 96384373-96384373 on Build GRCh38
rs953787101 Chr.11: 96384279-96384279 on Build GRCh38
rs778264160 Chr.11: 96384343-96384343 on Build GRCh38
rs75857759 Chr.11: 96384426-96384426 on Build GRCh38
rs771916792 Chr.11: 96384352-96384353 on Build GRCh38
rs994351966 Chr.11: 96384299-96384299 on Build GRCh38
rs769798181 Chr.11: 96384381-96384381 on Build GRCh38
rs774545821 Chr.11: 96384433-96384433 on Build GRCh38
rs369937047 Chr.11: 96384273-96384273 on Build GRCh38
rs776541128 Chr.11: 96384298-96384298 on Build GRCh38
rs779657939 Chr.11: 96384362-96384362 on Build GRCh38
rs773453775 Chr.11: 96384360-96384360 on Build GRCh38
rs767218465 Chr.11: 96384378-96384378 on Build GRCh38
rs1018392735 Chr.11: 96384323-96384323 on Build GRCh38
rs771651301 Chr.11: 96384283-96384283 on Build GRCh38
rs377425464 Chr.11: 96384421-96384421 on Build GRCh38
rs888645984 Chr.11: 96384316-96384316 on Build GRCh38
rs144935548 Chr.11: 96384287-96384287 on Build GRCh38
rs575911765 Chr.11: 96384409-96384409 on Build GRCh38
rs761646107 Chr.11: 96384300-96384300 on Build GRCh38
rs533065669 Chr.11: 96384319-96384319 on Build GRCh38
rs556503109 Chr.11: 96384379-96384379 on Build GRCh38
rs768309826 Chr.11: 96384293-96384293 on Build GRCh38
rs200083008 Chr.11: 96384277-96384277 on Build GRCh38
rs143022124 Chr.11: 96384356-96384356 on Build GRCh38
rs17851661 Chr.11: 96384303-96384303 on Build GRCh38
rs3748261 Chr.11: 96384365-96384365 on Build GRCh38
rs749883135 Chr.11: 96384354-96384354 on Build GRCh38
rs779699828 Chr.11: 96384285-96384285 on Build GRCh38
rs373116078 Chr.11: 96384336-96384336 on Build GRCh38
rs748628798 Chr.11: 96384330-96384330 on Build GRCh38
rs753170761 Chr.11: 96384335-96384335 on Build GRCh38
rs777330495 Chr.11: 96384306-96384306 on Build GRCh38
rs1006118849 Chr.11: 96384317-96384317 on Build GRCh38
rs767757717 Chr.11: 96384331-96384331 on Build GRCh38
rs778387124 Chr.11: 96384274-96384274 on Build GRCh38
rs150358447 Chr.11: 96384266-96384266 on Build GRCh38

View Full Product Details