Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17524095-17524360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR2
Entrez Gene ID
27443
Gene Name
CECR2, histone acetyl-lysine reader
Gene Aliases
-
UniGene
Hs.231895
Gene Chromosome Location
Chr.22: 17359949-17558155 on Build GRCh38
Transcripts
NM_001290046.1 NM_001290047.1 XM_011546128.2 XM_011546129.2 XM_011546132.2 XM_017028785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448046 c.848T>C p.L283P Chr.22: 17524221-17524221 on Build GRCh38
5513664 c.865C>T p.R289C Chr.22: 17524238-17524238 on Build GRCh38
4447751 c.793G>A p.E265K Chr.22: 17524166-17524166 on Build GRCh38
4448045 c.992T>C p.L331P Chr.22: 17524221-17524221 on Build GRCh38
4602579 c.917A>T p.K306M Chr.22: 17524203-17524203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749069125 Chr.22: 17524187-17524187 on Build GRCh38
rs929812794 Chr.22: 17524282-17524282 on Build GRCh38
rs767550931 Chr.22: 17524323-17524323 on Build GRCh38
rs138101258 Chr.22: 17524155-17524155 on Build GRCh38
rs900283274 Chr.22: 17524156-17524156 on Build GRCh38
rs767386996 Chr.22: 17524333-17524334 on Build GRCh38
rs777876426 Chr.22: 17524288-17524288 on Build GRCh38
rs750620736 Chr.22: 17524277-17524277 on Build GRCh38
rs766620277 Chr.22: 17524143-17524143 on Build GRCh38
rs754787038 Chr.22: 17524279-17524279 on Build GRCh38
rs751586411 Chr.22: 17524252-17524252 on Build GRCh38
rs770353990 Chr.22: 17524295-17524295 on Build GRCh38
rs368613674 Chr.22: 17524286-17524286 on Build GRCh38
rs375042326 Chr.22: 17524297-17524297 on Build GRCh38
rs778972186 Chr.22: 17524317-17524317 on Build GRCh38
rs764220976 Chr.22: 17524160-17524160 on Build GRCh38
rs760846394 Chr.22: 17524326-17524326 on Build GRCh38
rs774841529 Chr.22: 17524321-17524321 on Build GRCh38
rs368031555 Chr.22: 17524307-17524307 on Build GRCh38
rs770194958 Chr.22: 17524308-17524308 on Build GRCh38
rs748152983 Chr.22: 17524219-17524219 on Build GRCh38
rs541330262 Chr.22: 17524275-17524276 on Build GRCh38
rs374499391 Chr.22: 17524276-17524276 on Build GRCh38
rs778851814 Chr.22: 17524209-17524209 on Build GRCh38
rs781288775 Chr.22: 17524183-17524183 on Build GRCh38
rs73159550 Chr.22: 17524315-17524315 on Build GRCh38
rs765100621 Chr.22: 17524335-17524335 on Build GRCh38
rs751780457 Chr.22: 17524162-17524162 on Build GRCh38
rs759879618 Chr.22: 17524240-17524240 on Build GRCh38
rs536063005 Chr.22: 17524241-17524241 on Build GRCh38
rs964732053 Chr.22: 17524139-17524139 on Build GRCh38
rs996241613 Chr.22: 17524319-17524319 on Build GRCh38
rs868822830 Chr.22: 17524218-17524218 on Build GRCh38
rs757033889 Chr.22: 17524302-17524314 on Build GRCh38
rs771704119 Chr.22: 17524220-17524220 on Build GRCh38
rs375892327 Chr.22: 17524322-17524322 on Build GRCh38
rs367584284 Chr.22: 17524182-17524182 on Build GRCh38
rs771720298 Chr.22: 17524118-17524118 on Build GRCh38
rs772888133 Chr.22: 17524222-17524222 on Build GRCh38
rs73389185 Chr.22: 17524287-17524287 on Build GRCh38
rs557048827 Chr.22: 17524228-17524228 on Build GRCh38
rs777102530 Chr.22: 17524145-17524145 on Build GRCh38
rs370462260 Chr.22: 17524154-17524154 on Build GRCh38
rs573791910 Chr.22: 17524239-17524239 on Build GRCh38
rs746478829 Chr.22: 17524289-17524289 on Build GRCh38
rs764131143 Chr.22: 17524249-17524249 on Build GRCh38
rs955556056 Chr.22: 17524202-17524203 on Build GRCh38
rs753210062 Chr.22: 17524157-17524157 on Build GRCh38
rs377543712 Chr.22: 17524120-17524120 on Build GRCh38
rs750651769 Chr.22: 17524324-17524324 on Build GRCh38
rs923483211 Chr.22: 17524169-17524169 on Build GRCh38
rs754730786 Chr.22: 17524202-17524202 on Build GRCh38
rs1046176641 Chr.22: 17524258-17524258 on Build GRCh38
rs924189810 Chr.22: 17524210-17524210 on Build GRCh38
rs775966102 Chr.22: 17524242-17524242 on Build GRCh38
rs758418427 Chr.22: 17524158-17524158 on Build GRCh38
rs371557341 Chr.22: 17524244-17524244 on Build GRCh38
rs750440974 Chr.22: 17524115-17524115 on Build GRCh38
rs368913946 Chr.22: 17524325-17524325 on Build GRCh38
rs770569105 Chr.22: 17524238-17524238 on Build GRCh38
rs757310038 Chr.22: 17524269-17524269 on Build GRCh38

View Full Product Details