Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35264988-35265255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGXB4
Entrez Gene ID
10042
Gene Name
HMG-box containing 4
Gene Aliases
HMG2L1, HMGBCG, THC211630
UniGene
Hs.588815
Gene Chromosome Location
Chr.22: 35257274-35295807 on Build GRCh38
Transcripts
NM_001003681.2 NR_027780.1 XM_006724100.3 XM_006724101.3 XM_006724102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281732 c.685C>T p.R229W Chr.22: 35265073-35265073 on Build GRCh38
4103746 c.796G>T p.G266C Chr.22: 35265184-35265184 on Build GRCh38
4103745 c.780A>G p.S260S Chr.22: 35265168-35265168 on Build GRCh38
5742103 c.623A>G p.E208G Chr.22: 35265011-35265011 on Build GRCh38
7130381 c.803G>A p.G268E Chr.22: 35265191-35265191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777204741 Chr.22: 35265181-35265181 on Build GRCh38
rs753726655 Chr.22: 35265223-35265223 on Build GRCh38
rs538821671 Chr.22: 35265073-35265073 on Build GRCh38
rs749270276 Chr.22: 35265077-35265077 on Build GRCh38
rs1038770016 Chr.22: 35265224-35265224 on Build GRCh38
rs750301294 Chr.22: 35265070-35265070 on Build GRCh38
rs759885097 Chr.22: 35265044-35265044 on Build GRCh38
rs750687808 Chr.22: 35265147-35265147 on Build GRCh38
rs375704381 Chr.22: 35265083-35265083 on Build GRCh38
rs751647131 Chr.22: 35265060-35265060 on Build GRCh38
rs747728307 Chr.22: 35265172-35265172 on Build GRCh38
rs1030579074 Chr.22: 35265036-35265036 on Build GRCh38
rs749592645 Chr.22: 35265190-35265190 on Build GRCh38
rs769262061 Chr.22: 35265192-35265192 on Build GRCh38
rs111507753 Chr.22: 35265025-35265025 on Build GRCh38
rs779656431 Chr.22: 35265074-35265074 on Build GRCh38
rs760999697 Chr.22: 35265211-35265211 on Build GRCh38
rs540103611 Chr.22: 35265230-35265230 on Build GRCh38
rs561286969 Chr.22: 35265199-35265199 on Build GRCh38
rs758222733 Chr.22: 35265170-35265172 on Build GRCh38
rs771684284 Chr.22: 35265033-35265033 on Build GRCh38
rs199921358 Chr.22: 35265134-35265134 on Build GRCh38
rs576892349 Chr.22: 35265067-35265067 on Build GRCh38
rs770448105 Chr.22: 35265183-35265183 on Build GRCh38
rs764948707 Chr.22: 35265231-35265231 on Build GRCh38
rs75473111 Chr.22: 35265008-35265008 on Build GRCh38
rs764254582 Chr.22: 35265054-35265054 on Build GRCh38
rs752641529 Chr.22: 35265232-35265232 on Build GRCh38
rs778360774 Chr.22: 35265187-35265187 on Build GRCh38
rs370732199 Chr.22: 35265168-35265168 on Build GRCh38
rs972108898 Chr.22: 35265024-35265024 on Build GRCh38
rs139836644 Chr.22: 35265066-35265066 on Build GRCh38
rs370541896 Chr.22: 35265082-35265082 on Build GRCh38
rs574478294 Chr.22: 35265219-35265219 on Build GRCh38
rs917796660 Chr.22: 35265156-35265156 on Build GRCh38
rs761485667 Chr.22: 35265030-35265030 on Build GRCh38
rs764475927 Chr.22: 35265126-35265126 on Build GRCh38
rs1041394688 Chr.22: 35265107-35265107 on Build GRCh38
rs746594486 Chr.22: 35265102-35265102 on Build GRCh38
rs752967716 Chr.22: 35265047-35265047 on Build GRCh38
rs773686909 Chr.22: 35265028-35265028 on Build GRCh38
rs1057388836 Chr.22: 35265184-35265184 on Build GRCh38
rs777560353 Chr.22: 35265213-35265214 on Build GRCh38
rs145147733 Chr.22: 35265182-35265182 on Build GRCh38
rs761971332 Chr.22: 35265136-35265136 on Build GRCh38
rs765734557 Chr.22: 35265046-35265046 on Build GRCh38
rs766230175 Chr.22: 35265159-35265159 on Build GRCh38
rs775665083 Chr.22: 35265117-35265117 on Build GRCh38
rs756241387 Chr.22: 35265154-35265154 on Build GRCh38
rs146646334 Chr.22: 35265094-35265094 on Build GRCh38
rs747637071 Chr.22: 35265091-35265091 on Build GRCh38
rs141408412 Chr.22: 35265146-35265146 on Build GRCh38
rs773385936 Chr.22: 35265210-35265210 on Build GRCh38
rs368621812 Chr.22: 35265104-35265104 on Build GRCh38
rs752708702 Chr.22: 35265010-35265012 on Build GRCh38
rs745507094 Chr.22: 35265198-35265198 on Build GRCh38
rs763543566 Chr.22: 35265123-35265123 on Build GRCh38
rs767728221 Chr.22: 35265207-35265207 on Build GRCh38
rs753547991 Chr.22: 35265162-35265162 on Build GRCh38
rs759249598 Chr.22: 35265229-35265229 on Build GRCh38
rs757911397 Chr.22: 35265180-35265180 on Build GRCh38
rs150944787 Chr.22: 35265041-35265041 on Build GRCh38

View Full Product Details