Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8072712-8072937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12B
Entrez Gene ID
242
Gene Name
arachidonate 12-lipoxygenase, 12R type
Gene Aliases
12R-LOX, ARCI2
UniGene
Hs.136574
Gene Chromosome Location
Chr.17: 8072636-8087703 on Build GRCh38
Transcripts
NM_001139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6973710 c.1976G>A p.R659Q Chr.17: 8072901-8072901 on Build GRCh38
985921 c.1959C>T p.F653F Chr.17: 8072918-8072918 on Build GRCh38
4386504 c.2003G>A p.R668H Chr.17: 8072874-8072874 on Build GRCh38
6598664 c.2022C>T p.H674H Chr.17: 8072855-8072855 on Build GRCh38
985919 c.1989G>T p.E663D Chr.17: 8072888-8072888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758255070 Chr.17: 8072894-8072894 on Build GRCh38
rs778801170 Chr.17: 8072760-8072760 on Build GRCh38
rs550272024 Chr.17: 8072855-8072855 on Build GRCh38
rs749332506 Chr.17: 8072906-8072906 on Build GRCh38
rs774876393 Chr.17: 8072830-8072830 on Build GRCh38
rs745867054 Chr.17: 8072892-8072892 on Build GRCh38
rs772035116 Chr.17: 8072766-8072766 on Build GRCh38
rs748377475 Chr.17: 8072795-8072795 on Build GRCh38
rs143164748 Chr.17: 8072822-8072822 on Build GRCh38
rs763381583 Chr.17: 8072750-8072750 on Build GRCh38
rs756877833 Chr.17: 8072874-8072874 on Build GRCh38
rs766901670 Chr.17: 8072753-8072753 on Build GRCh38
rs779729947 Chr.17: 8072901-8072901 on Build GRCh38
rs751607606 Chr.17: 8072846-8072847 on Build GRCh38
rs373588263 Chr.17: 8072764-8072764 on Build GRCh38
rs367750142 Chr.17: 8072789-8072789 on Build GRCh38
rs760052556 Chr.17: 8072808-8072808 on Build GRCh38
rs773173020 Chr.17: 8072815-8072815 on Build GRCh38
rs763413541 Chr.17: 8072819-8072819 on Build GRCh38
rs773297344 Chr.17: 8072905-8072905 on Build GRCh38
rs769857850 Chr.17: 8072806-8072806 on Build GRCh38
rs748339571 Chr.17: 8072904-8072904 on Build GRCh38
rs776908057 Chr.17: 8072793-8072793 on Build GRCh38
rs397514528 Chr.17: 8072841-8072841 on Build GRCh38
rs771062878 Chr.17: 8072907-8072907 on Build GRCh38
rs147489840 Chr.17: 8072868-8072868 on Build GRCh38
rs772504655 Chr.17: 8072846-8072846 on Build GRCh38
rs774823007 Chr.17: 8072908-8072908 on Build GRCh38
rs778964875 Chr.17: 8072878-8072878 on Build GRCh38
rs993532550 Chr.17: 8072767-8072767 on Build GRCh38
rs759885347 Chr.17: 8072832-8072832 on Build GRCh38

View Full Product Details