Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196219820-196220093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4764276 c.4388A>G p.N1463S Chr.2: 196220059-196220059 on Build GRCh38
7185539 c.4393A>C p.T1465P Chr.2: 196220054-196220054 on Build GRCh38
320794 c.4369G>A p.D1457N Chr.2: 196220078-196220078 on Build GRCh38
5585411 c.4383G>A p.W1461* Chr.2: 196220064-196220064 on Build GRCh38
4912298 c.4375A>G p.S1459G Chr.2: 196220072-196220072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753312009 Chr.2: 196220067-196220067 on Build GRCh38
rs763162697 Chr.2: 196220052-196220053 on Build GRCh38
rs1049256713 Chr.2: 196219941-196219941 on Build GRCh38
rs995425895 Chr.2: 196219917-196219917 on Build GRCh38
rs1531114 Chr.2: 196219871-196219871 on Build GRCh38
rs777216163 Chr.2: 196219859-196219859 on Build GRCh38
rs759875255 Chr.2: 196220034-196220034 on Build GRCh38
rs565418326 Chr.2: 196219978-196219978 on Build GRCh38
rs568799962 Chr.2: 196219945-196219945 on Build GRCh38
rs112491032 Chr.2: 196219948-196219948 on Build GRCh38
rs543315119 Chr.2: 196219884-196219884 on Build GRCh38
rs545527605 Chr.2: 196219967-196219967 on Build GRCh38
rs1029264052 Chr.2: 196219888-196219888 on Build GRCh38
rs765511221 Chr.2: 196220046-196220046 on Build GRCh38
rs1531113 Chr.2: 196219863-196219863 on Build GRCh38
rs756975748 Chr.2: 196219969-196219969 on Build GRCh38
rs376196967 Chr.2: 196219964-196219964 on Build GRCh38
rs184161183 Chr.2: 196219877-196219877 on Build GRCh38
rs766579547 Chr.2: 196219991-196219991 on Build GRCh38
rs201066286 Chr.2: 196220029-196220029 on Build GRCh38
rs762399108 Chr.2: 196219903-196219903 on Build GRCh38
rs527782139 Chr.2: 196220017-196220017 on Build GRCh38
rs767052627 Chr.2: 196219984-196219984 on Build GRCh38

View Full Product Details